Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 22625415-22625579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCF
Entrez Gene ID
2188
Gene Name
Fanconi anemia complementation group F
Gene Aliases
FAF
UniGene
Hs.632151
Gene Chromosome Location
Chr.11: 22622533-22625841 on Build GRCh38
Transcripts
NM_022725.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5019891 c.387C>T p.L129L Chr.11: 22625424-22625424 on Build GRCh38
7367580 c.350C>T p.P117L Chr.11: 22625461-22625461 on Build GRCh38
4990144 c.314A>G p.D105G Chr.11: 22625497-22625497 on Build GRCh38
6591433 c.270C>T p.D90D Chr.11: 22625541-22625541 on Build GRCh38
5443135 c.373G>A p.D125N Chr.11: 22625438-22625438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746415789 Chr.11: 22625542-22625542 on Build GRCh38
rs374572943 Chr.11: 22625461-22625461 on Build GRCh38
rs377565368 Chr.11: 22625533-22625533 on Build GRCh38
rs776347203 Chr.11: 22625522-22625522 on Build GRCh38
rs200874455 Chr.11: 22625458-22625458 on Build GRCh38
rs781455843 Chr.11: 22625481-22625481 on Build GRCh38
rs552500538 Chr.11: 22625541-22625541 on Build GRCh38
rs761672325 Chr.11: 22625523-22625523 on Build GRCh38
rs104894222 Chr.11: 22625484-22625484 on Build GRCh38
rs61752920 Chr.11: 22625438-22625438 on Build GRCh38
rs757993614 Chr.11: 22625473-22625473 on Build GRCh38
rs199578614 Chr.11: 22625532-22625532 on Build GRCh38
rs767379904 Chr.11: 22625450-22625450 on Build GRCh38
rs760472320 Chr.11: 22625455-22625455 on Build GRCh38
rs199705194 Chr.11: 22625439-22625439 on Build GRCh38
rs985046037 Chr.11: 22625496-22625496 on Build GRCh38
rs760571335 Chr.11: 22625510-22625510 on Build GRCh38
rs752081138 Chr.11: 22625548-22625548 on Build GRCh38
rs759470099 Chr.11: 22625447-22625447 on Build GRCh38
rs886048158 Chr.11: 22625548-22625550 on Build GRCh38
rs779538319 Chr.11: 22625469-22625469 on Build GRCh38
rs749311845 Chr.11: 22625492-22625492 on Build GRCh38
rs201357673 Chr.11: 22625441-22625441 on Build GRCh38
rs750121502 Chr.11: 22625525-22625525 on Build GRCh38
rs771023850 Chr.11: 22625493-22625493 on Build GRCh38
rs775367657 Chr.11: 22625503-22625503 on Build GRCh38
rs755335784 Chr.11: 22625480-22625480 on Build GRCh38
rs730880277 Chr.11: 22625559-22625581 on Build GRCh38
rs769892426 Chr.11: 22625444-22625444 on Build GRCh38
rs749884571 Chr.11: 22625466-22625466 on Build GRCh38
rs773334179 Chr.11: 22625446-22625446 on Build GRCh38
rs587778341 Chr.11: 22625555-22625555 on Build GRCh38
rs370200638 Chr.11: 22625543-22625543 on Build GRCh38
rs763928544 Chr.11: 22625511-22625511 on Build GRCh38
rs200995539 Chr.11: 22625489-22625489 on Build GRCh38
rs757554175 Chr.11: 22625535-22625535 on Build GRCh38
rs780657159 Chr.11: 22625474-22625474 on Build GRCh38
rs372625322 Chr.11: 22625462-22625462 on Build GRCh38
rs373385251 Chr.11: 22625479-22625479 on Build GRCh38
rs149490050 Chr.11: 22625460-22625460 on Build GRCh38
rs775422716 Chr.11: 22625451-22625451 on Build GRCh38
rs764924585 Chr.11: 22625524-22625524 on Build GRCh38
rs146352365 Chr.11: 22625553-22625553 on Build GRCh38
rs773430381 Chr.11: 22625491-22625491 on Build GRCh38

View Full Product Details