Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 26382838-26382993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK8
Entrez Gene ID
1024
Gene Name
cyclin dependent kinase 8
Gene Aliases
K35
UniGene
Hs.382306
Gene Chromosome Location
Chr.13: 26254104-26405238 on Build GRCh38
Transcripts
NM_001260.2 NM_001318368.1 XM_005266216.4 XM_011534864.1 XM_011534865.2 XM_017020317.1 XM_017020318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5342311 c.496C>T p.R166* Chr.13: 26382853-26382853 on Build GRCh38
70127 c.493G>C p.E165Q Chr.13: 26382850-26382850 on Build GRCh38
5528501 c.490C>T p.P164S Chr.13: 26382847-26382847 on Build GRCh38
1366128 c.489T>C p.G163G Chr.13: 26382846-26382846 on Build GRCh38
6138870 c.514G>A p.A172T Chr.13: 26382871-26382871 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1009490631 Chr.13: 26382907-26382907 on Build GRCh38
rs759478064 Chr.13: 26382888-26382888 on Build GRCh38
rs781129176 Chr.13: 26382913-26382913 on Build GRCh38
rs770570887 Chr.13: 26382891-26382896 on Build GRCh38
rs544120349 Chr.13: 26382902-26382902 on Build GRCh38
rs765422516 Chr.13: 26382896-26382896 on Build GRCh38
rs750174273 Chr.13: 26382921-26382921 on Build GRCh38
rs1020343364 Chr.13: 26382911-26382911 on Build GRCh38
rs958738811 Chr.13: 26382953-26382953 on Build GRCh38
rs973261589 Chr.13: 26382962-26382962 on Build GRCh38
rs991363962 Chr.13: 26382947-26382947 on Build GRCh38
rs1035324698 Chr.13: 26382922-26382922 on Build GRCh38
rs748403142 Chr.13: 26382873-26382877 on Build GRCh38
rs757268110 Chr.13: 26382912-26382912 on Build GRCh38
rs963657465 Chr.13: 26382960-26382960 on Build GRCh38
rs116597905 Chr.13: 26382943-26382943 on Build GRCh38
rs547285437 Chr.13: 26382959-26382959 on Build GRCh38
rs369035839 Chr.13: 26382901-26382901 on Build GRCh38
rs915836904 Chr.13: 26382950-26382950 on Build GRCh38

View Full Product Details