Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43532559-43532717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX8
Entrez Gene ID
1659
Gene Name
DEAH-box helicase 8
Gene Aliases
DDX8, HRH1, PRP22, PRPF22
UniGene
Hs.463105
Gene Chromosome Location
Chr.17: 43483865-43544787 on Build GRCh38
Transcripts
NM_001322219.1 NM_001079675.2 NM_001261437.1 NM_001261438.1 NM_001986.2 XM_011524514.1 XM_017024349.1

Gene Symbol
ETV4
Entrez Gene ID
2118
Gene Name
ETS variant 4
Gene Aliases
E1A-F, E1AF, PEA3, PEAS3
UniGene
Hs.434059
Gene Chromosome Location
Chr.17: 43527843-43546432 on Build GRCh38
Transcripts
NM_001322219.1 NM_001079675.2 NM_001261437.1 NM_001261438.1 NM_001986.2 XM_011524514.1 XM_017024349.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3517932 c.792G>A p.T264T Chr.17: 43532693-43532693 on Build GRCh38
4130117 c.770T>G p.V257G Chr.17: 43532715-43532715 on Build GRCh38
3795656 c.777C>T p.I259I Chr.17: 43532708-43532708 on Build GRCh38
5624352 c.792G>T p.T264T Chr.17: 43532693-43532693 on Build GRCh38
4559461 c.805G>A p.D269N Chr.17: 43532680-43532680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749666909 Chr.17: 43532686-43532686 on Build GRCh38
rs904612883 Chr.17: 43532622-43532622 on Build GRCh38
rs770730900 Chr.17: 43532680-43532680 on Build GRCh38
rs113285687 Chr.17: 43532673-43532673 on Build GRCh38
rs935419928 Chr.17: 43532582-43532582 on Build GRCh38
rs142548326 Chr.17: 43532694-43532694 on Build GRCh38
rs1040460384 Chr.17: 43532595-43532595 on Build GRCh38
rs746622168 Chr.17: 43532675-43532675 on Build GRCh38
rs901395941 Chr.17: 43532646-43532646 on Build GRCh38
rs536689944 Chr.17: 43532663-43532664 on Build GRCh38
rs201905867 Chr.17: 43532628-43532628 on Build GRCh38
rs760544272 Chr.17: 43532650-43532650 on Build GRCh38
rs769039480 Chr.17: 43532687-43532687 on Build GRCh38
rs774978942 Chr.17: 43532693-43532693 on Build GRCh38
rs777537367 Chr.17: 43532664-43532664 on Build GRCh38
rs780509931 Chr.17: 43532681-43532681 on Build GRCh38
rs764704828 Chr.17: 43532665-43532668 on Build GRCh38
rs752137870 Chr.17: 43532633-43532633 on Build GRCh38
rs557779520 Chr.17: 43532635-43532635 on Build GRCh38
rs998466217 Chr.17: 43532671-43532671 on Build GRCh38
rs1043157718 Chr.17: 43532607-43532607 on Build GRCh38

View Full Product Details