Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48792263-48792437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA1
Entrez Gene ID
2623
Gene Name
GATA binding protein 1
Gene Aliases
ERYF1, GATA-1, GF-1, GF1, NF-E1, NFE1, XLANP, XLTDA, XLTT
UniGene
Hs.765
Gene Chromosome Location
Chr.X: 48786540-48794311 on Build GRCh38
Transcripts
NM_002049.3 XM_011543897.2 XM_011543898.2

SNP Detail

SNP ID Chromosome Location
rs782127979 Chr.X: 48792302-48792303 on Build GRCh38
rs782273718 Chr.X: 48792286-48792286 on Build GRCh38
rs373414851 Chr.X: 48792374-48792374 on Build GRCh38
rs782039463 Chr.X: 48792352-48792352 on Build GRCh38
rs387907207 Chr.X: 48792370-48792370 on Build GRCh38
rs782435835 Chr.X: 48792292-48792292 on Build GRCh38
rs782347891 Chr.X: 48792313-48792313 on Build GRCh38
rs587776454 Chr.X: 48792346-48792346 on Build GRCh38
rs104894815 Chr.X: 48792337-48792337 on Build GRCh38
rs782785151 Chr.X: 48792345-48792345 on Build GRCh38
rs782282009 Chr.X: 48792387-48792387 on Build GRCh38
rs104894809 Chr.X: 48792371-48792371 on Build GRCh38
rs104894816 Chr.X: 48792377-48792377 on Build GRCh38
rs137852312 Chr.X: 48792346-48792347 on Build GRCh38
rs782068997 Chr.X: 48792301-48792301 on Build GRCh38
rs781895544 Chr.X: 48792348-48792348 on Build GRCh38
rs104894808 Chr.X: 48792376-48792376 on Build GRCh38
rs369454016 Chr.X: 48792298-48792298 on Build GRCh38
rs376394957 Chr.X: 48792384-48792384 on Build GRCh38
rs370522986 Chr.X: 48792402-48792402 on Build GRCh38

View Full Product Details