Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 67545415-67545547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AR
Entrez Gene ID
367
Gene Name
androgen receptor
Gene Aliases
AIS, AR8, DHTR, HUMARA, HYSP1, KD, NR3C4, SBMA, SMAX1, TFM
UniGene
Hs.76704
Gene Chromosome Location
Chr.X: 67544032-67730619 on Build GRCh38
Transcripts
NM_000044.3

SNP Detail

SNP ID Chromosome Location
rs749864595 Chr.X: 67545508-67545508 on Build GRCh38
rs781422644 Chr.X: 67545457-67545457 on Build GRCh38
rs746181830 Chr.X: 67545458-67545458 on Build GRCh38
rs200817861 Chr.X: 67545488-67545488 on Build GRCh38
rs868474062 Chr.X: 67545493-67545493 on Build GRCh38
rs772890200 Chr.X: 67545521-67545521 on Build GRCh38
rs147677446 Chr.X: 67545470-67545470 on Build GRCh38
rs751756072 Chr.X: 67545446-67545446 on Build GRCh38
rs375624067 Chr.X: 67545479-67545479 on Build GRCh38
rs1031938966 Chr.X: 67545527-67545527 on Build GRCh38
rs771463719 Chr.X: 67545513-67545513 on Build GRCh38
rs777689173 Chr.X: 67545448-67545448 on Build GRCh38
rs976306779 Chr.X: 67545492-67545492 on Build GRCh38
rs777886419 Chr.X: 67545437-67545437 on Build GRCh38

View Full Product Details