Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 64190288-64190458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMER1
Entrez Gene ID
139285
Gene Name
APC membrane recruitment protein 1
Gene Aliases
FAM123B, OSCS, WTX
UniGene
Hs.314225
Gene Chromosome Location
Chr.X: 64185117-64205744 on Build GRCh38
Transcripts
NM_152424.3

SNP Detail

SNP ID Chromosome Location
rs745847898 Chr.X: 64190351-64190351 on Build GRCh38
rs370290063 Chr.X: 64190418-64190418 on Build GRCh38
rs968815543 Chr.X: 64190388-64190388 on Build GRCh38
rs374610712 Chr.X: 64190430-64190430 on Build GRCh38
rs745398718 Chr.X: 64190372-64190372 on Build GRCh38
rs763440022 Chr.X: 64190341-64190341 on Build GRCh38
rs751641117 Chr.X: 64190407-64190407 on Build GRCh38
rs768563880 Chr.X: 64190433-64190433 on Build GRCh38
rs770055483 Chr.X: 64190330-64190330 on Build GRCh38
rs769565705 Chr.X: 64190398-64190398 on Build GRCh38
rs775915295 Chr.X: 64190337-64190337 on Build GRCh38
rs758378585 Chr.X: 64190400-64190400 on Build GRCh38
rs778708592 Chr.X: 64190317-64190317 on Build GRCh38
rs376353035 Chr.X: 64190344-64190344 on Build GRCh38
rs753464604 Chr.X: 64190382-64190382 on Build GRCh38
rs772682408 Chr.X: 64190358-64190358 on Build GRCh38
rs746088303 Chr.X: 64190428-64190428 on Build GRCh38
rs780395189 Chr.X: 64190432-64190432 on Build GRCh38
rs374729339 Chr.X: 64190390-64190390 on Build GRCh38
rs777905301 Chr.X: 64190403-64190403 on Build GRCh38
rs371014578 Chr.X: 64190376-64190376 on Build GRCh38
rs529181603 Chr.X: 64190350-64190350 on Build GRCh38
rs376943508 Chr.X: 64190394-64190394 on Build GRCh38
rs924025276 Chr.X: 64190334-64190335 on Build GRCh38
rs746199690 Chr.X: 64190322-64190322 on Build GRCh38
rs966101634 Chr.X: 64190425-64190425 on Build GRCh38
rs766008251 Chr.X: 64190380-64190380 on Build GRCh38
rs867885720 Chr.X: 64190339-64190339 on Build GRCh38
rs780015689 Chr.X: 64190413-64190413 on Build GRCh38

View Full Product Details