Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 13926508-13926646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC4
Entrez Gene ID
2072
Gene Name
ERCC excision repair 4, endonuclease catalytic subunit
Gene Aliases
ERCC11, FANCQ, RAD1, XFEPS, XPF
UniGene
Hs.567265
Gene Chromosome Location
Chr.16: 13920138-13952348 on Build GRCh38
Transcripts
NM_005236.2 XM_011522424.2 XM_017023043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5516728 c.457C>T p.R153C Chr.16: 13926629-13926629 on Build GRCh38
6953060 c.406G>A p.A136T Chr.16: 13926578-13926578 on Build GRCh38
6917294 c.458G>C p.R153P Chr.16: 13926630-13926630 on Build GRCh38
6985212 c.467A>G p.N156S Chr.16: 13926639-13926639 on Build GRCh38
4440429 c.389-9delC p.? Chr.16: 13926552-13926552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775970324 Chr.16: 13926580-13926580 on Build GRCh38
rs748894431 Chr.16: 13926569-13926569 on Build GRCh38
rs139989614 Chr.16: 13926619-13926619 on Build GRCh38
rs772979483 Chr.16: 13926567-13926567 on Build GRCh38
rs746763106 Chr.16: 13926545-13926545 on Build GRCh38
rs1002918079 Chr.16: 13926595-13926595 on Build GRCh38
rs145402255 Chr.16: 13926620-13926620 on Build GRCh38
rs147455220 Chr.16: 13926598-13926598 on Build GRCh38
rs769186966 Chr.16: 13926563-13926563 on Build GRCh38
rs765509563 Chr.16: 13926600-13926600 on Build GRCh38
rs377014538 Chr.16: 13926621-13926621 on Build GRCh38
rs772606808 Chr.16: 13926576-13926576 on Build GRCh38
rs774717152 Chr.16: 13926571-13926571 on Build GRCh38
rs138724289 Chr.16: 13926592-13926592 on Build GRCh38
rs760865964 Chr.16: 13926593-13926593 on Build GRCh38
rs369626998 Chr.16: 13926552-13926552 on Build GRCh38
rs764093404 Chr.16: 13926597-13926597 on Build GRCh38
rs377224276 Chr.16: 13926556-13926556 on Build GRCh38
rs781256638 Chr.16: 13926554-13926554 on Build GRCh38
rs745648039 Chr.16: 13926573-13926573 on Build GRCh38
rs751450497 Chr.16: 13926602-13926602 on Build GRCh38

View Full Product Details