Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 64971921-64972087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH11
Entrez Gene ID
1009
Gene Name
cadherin 11
Gene Aliases
CAD11, CDHOB, OB, OSF-4
UniGene
Hs.116471
Gene Chromosome Location
Chr.16: 64943753-65126112 on Build GRCh38
Transcripts
NM_001308392.1 NM_001797.3 XM_005255761.3 XM_005255762.2 XM_005255763.2 XM_011522803.2 XM_017022830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4061640 c.1460A>G p.K487R Chr.16: 64971995-64971995 on Build GRCh38
7295597 c.1449T>C p.D483D Chr.16: 64972006-64972006 on Build GRCh38
7295598 c.1449T>C p.D483D Chr.16: 64972006-64972006 on Build GRCh38
4966237 c.1460A>C p.K487T Chr.16: 64971995-64971995 on Build GRCh38
1324374 c.1510C>T p.P504S Chr.16: 64971945-64971945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148024624 Chr.16: 64972025-64972025 on Build GRCh38
rs370243280 Chr.16: 64972058-64972058 on Build GRCh38
rs762550510 Chr.16: 64971945-64971945 on Build GRCh38
rs781337558 Chr.16: 64971952-64971952 on Build GRCh38
rs761020632 Chr.16: 64972008-64972008 on Build GRCh38
rs534779962 Chr.16: 64971986-64971986 on Build GRCh38
rs767856649 Chr.16: 64971957-64971957 on Build GRCh38
rs80138971 Chr.16: 64972023-64972023 on Build GRCh38
rs411127 Chr.16: 64971970-64971970 on Build GRCh38
rs146549125 Chr.16: 64972009-64972009 on Build GRCh38
rs958989980 Chr.16: 64971955-64971955 on Build GRCh38
rs141063325 Chr.16: 64972031-64972031 on Build GRCh38
rs750853935 Chr.16: 64971981-64971981 on Build GRCh38
rs781127265 Chr.16: 64972059-64972059 on Build GRCh38
rs745658550 Chr.16: 64972063-64972063 on Build GRCh38
rs768156725 Chr.16: 64971949-64971949 on Build GRCh38
rs761504070 Chr.16: 64971956-64971956 on Build GRCh38
rs946153463 Chr.16: 64972010-64972010 on Build GRCh38
rs369963348 Chr.16: 64972052-64972052 on Build GRCh38
rs992858265 Chr.16: 64971985-64971985 on Build GRCh38
rs964186561 Chr.16: 64971944-64971944 on Build GRCh38

View Full Product Details