Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 61683663-61683787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRIP1
Entrez Gene ID
83990
Gene Name
BRCA1 interacting protein C-terminal helicase 1
Gene Aliases
BACH1, FANCJ, OF
UniGene
Hs.128903
Gene Chromosome Location
Chr.17: 61679186-61864120 on Build GRCh38
Transcripts
NM_032043.2 XM_011525332.2 XM_011525333.2 XM_011525334.2 XM_011525335.2 XM_011525336.2 XM_011525337.2 XM_011525338.2 XM_017025200.1 XM_017025201.1 XM_017025202.1 XM_017025203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
982262 c.3342G>A p.Q1114Q Chr.17: 61683704-61683704 on Build GRCh38
982260 c.3359A>C p.D1120A Chr.17: 61683687-61683687 on Build GRCh38
7076113 c.3376delG p.E1126fs*24 Chr.17: 61683670-61683670 on Build GRCh38
1563716 c.3266C>G p.S1089C Chr.17: 61683780-61683780 on Build GRCh38
982264 c.3289G>T p.E1097* Chr.17: 61683757-61683757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587781923 Chr.17: 61683748-61683748 on Build GRCh38
rs968860042 Chr.17: 61683712-61683712 on Build GRCh38
rs774105218 Chr.17: 61683761-61683761 on Build GRCh38
rs1034545913 Chr.17: 61683754-61683754 on Build GRCh38
rs781102464 Chr.17: 61683738-61683738 on Build GRCh38
rs111943191 Chr.17: 61683730-61683730 on Build GRCh38
rs369843642 Chr.17: 61683710-61683710 on Build GRCh38
rs748140041 Chr.17: 61683741-61683741 on Build GRCh38
rs772709195 Chr.17: 61683750-61683750 on Build GRCh38
rs757427210 Chr.17: 61683692-61683692 on Build GRCh38
rs748928248 Chr.17: 61683752-61683752 on Build GRCh38
rs876658746 Chr.17: 61683756-61683756 on Build GRCh38
rs770509300 Chr.17: 61683757-61683757 on Build GRCh38
rs587780248 Chr.17: 61683715-61683715 on Build GRCh38
rs864622072 Chr.17: 61683747-61683747 on Build GRCh38
rs779860140 Chr.17: 61683697-61683697 on Build GRCh38

View Full Product Details