Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 64947874-64948038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH11
Entrez Gene ID
1009
Gene Name
cadherin 11
Gene Aliases
CAD11, CDHOB, OB, OSF-4
UniGene
Hs.116471
Gene Chromosome Location
Chr.16: 64943753-65126112 on Build GRCh38
Transcripts
NM_001308392.1 NM_001797.3 XM_005255761.3 XM_005255762.2 XM_005255763.2 XM_011522803.2 XM_017022830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
471899 c.2117C>T p.P706L Chr.16: 64947877-64947877 on Build GRCh38
2993365 c.2029G>A p.D677N Chr.16: 64947965-64947965 on Build GRCh38
6079570 c.2082A>G p.K694K Chr.16: 64947912-64947912 on Build GRCh38
3510759 c.2040C>T p.T680T Chr.16: 64947954-64947954 on Build GRCh38
4972979 c.1966G>A p.V656I Chr.16: 64948028-64948028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757580985 Chr.16: 64947917-64947917 on Build GRCh38
rs777307769 Chr.16: 64947914-64947914 on Build GRCh38
rs879081628 Chr.16: 64947986-64947986 on Build GRCh38
rs765325404 Chr.16: 64947990-64947990 on Build GRCh38
rs878974367 Chr.16: 64947975-64947975 on Build GRCh38
rs776609272 Chr.16: 64947970-64947970 on Build GRCh38
rs768855337 Chr.16: 64947931-64947931 on Build GRCh38
rs751908119 Chr.16: 64947916-64947916 on Build GRCh38
rs780091829 Chr.16: 64947925-64947925 on Build GRCh38
rs759544481 Chr.16: 64947987-64947987 on Build GRCh38
rs764812249 Chr.16: 64947897-64947897 on Build GRCh38
rs35145978 Chr.16: 64947921-64947921 on Build GRCh38
rs866520799 Chr.16: 64947988-64947988 on Build GRCh38
rs746769837 Chr.16: 64947934-64947934 on Build GRCh38
rs746339300 Chr.16: 64947923-64947923 on Build GRCh38
rs752325229 Chr.16: 64947908-64947908 on Build GRCh38
rs371327463 Chr.16: 64947994-64947994 on Build GRCh38
rs774468321 Chr.16: 64947933-64947933 on Build GRCh38
rs186888660 Chr.16: 64947935-64947935 on Build GRCh38
rs749383194 Chr.16: 64947930-64947930 on Build GRCh38
rs758114163 Chr.16: 64947909-64947909 on Build GRCh38

View Full Product Details