Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58840342-58840497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAS
Entrez Gene ID
2778
Gene Name
GNAS complex locus
Gene Aliases
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
UniGene
Hs.125898
Gene Chromosome Location
Chr.20: 58839681-58911196 on Build GRCh38
Transcripts
NM_016592.3 XM_017027821.1 XM_017027822.1 NR_002785.2

Gene Symbol
GNAS-AS1
Entrez Gene ID
149775
Gene Name
GNAS antisense RNA 1
Gene Aliases
GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG
UniGene
Hs.122718
Gene Chromosome Location
Chr.20: 58818918-58850903 on Build GRCh38
Transcripts
NM_016592.3 XM_017027821.1 XM_017027822.1 NR_002785.2

SNP Detail

SNP ID Chromosome Location
rs575933995 Chr.20: 58840433-58840433 on Build GRCh38
rs769104795 Chr.20: 58840446-58840446 on Build GRCh38
rs753062074 Chr.20: 58840469-58840480 on Build GRCh38
rs747367482 Chr.20: 58840445-58840445 on Build GRCh38
rs753525197 Chr.20: 58840418-58840418 on Build GRCh38
rs373276011 Chr.20: 58840415-58840415 on Build GRCh38
rs771921968 Chr.20: 58840475-58840475 on Build GRCh38
rs201307445 Chr.20: 58840440-58840440 on Build GRCh38
rs951583745 Chr.20: 58840370-58840370 on Build GRCh38
rs760197051 Chr.20: 58840432-58840433 on Build GRCh38
rs766199286 Chr.20: 58840445-58840480 on Build GRCh38
rs752452409 Chr.20: 58840410-58840410 on Build GRCh38
rs748443274 Chr.20: 58840457-58840457 on Build GRCh38
rs369511579 Chr.20: 58840369-58840369 on Build GRCh38
rs190395960 Chr.20: 58840471-58840471 on Build GRCh38
rs756880829 Chr.20: 58840426-58840426 on Build GRCh38
rs587778387 Chr.20: 58840451-58840462 on Build GRCh38
rs1800902 Chr.20: 58840400-58840400 on Build GRCh38
rs762096209 Chr.20: 58840371-58840371 on Build GRCh38
rs936448388 Chr.20: 58840472-58840472 on Build GRCh38
rs758848179 Chr.20: 58840470-58840493 on Build GRCh38
rs563844600 Chr.20: 58840372-58840372 on Build GRCh38
rs587778388 Chr.20: 58840438-58840438 on Build GRCh38
rs773416371 Chr.20: 58840386-58840386 on Build GRCh38
rs755419386 Chr.20: 58840456-58840485 on Build GRCh38
rs779115304 Chr.20: 58840458-58840459 on Build GRCh38
rs781674491 Chr.20: 58840451-58840451 on Build GRCh38
rs543435005 Chr.20: 58840437-58840437 on Build GRCh38
rs199534645 Chr.20: 58840470-58840470 on Build GRCh38
rs775455733 Chr.20: 58840479-58840479 on Build GRCh38
rs942246524 Chr.20: 58840365-58840365 on Build GRCh38
rs144845382 Chr.20: 58840382-58840382 on Build GRCh38
rs770140447 Chr.20: 58840468-58840468 on Build GRCh38
rs375054629 Chr.20: 58840364-58840364 on Build GRCh38
rs148910090 Chr.20: 58840427-58840427 on Build GRCh38
rs759617960 Chr.20: 58840392-58840392 on Build GRCh38

View Full Product Details