Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 26254718-26254846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK8
Entrez Gene ID
1024
Gene Name
cyclin dependent kinase 8
Gene Aliases
K35
UniGene
Hs.382306
Gene Chromosome Location
Chr.13: 26254104-26405238 on Build GRCh38
Transcripts
NM_001260.2 NM_001318368.1 XM_011534864.1 XM_017020317.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3793222 c.102C>T p.H34H Chr.13: 26254743-26254743 on Build GRCh38
6625600 c.89G>A p.G30D Chr.13: 26254730-26254730 on Build GRCh38
4671186 c.128G>A p.G43E Chr.13: 26254769-26254769 on Build GRCh38
3738430 c.97G>A p.G33S Chr.13: 26254738-26254738 on Build GRCh38
5624253 c.95A>C p.Y32S Chr.13: 26254736-26254736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377345991 Chr.13: 26254818-26254818 on Build GRCh38
rs927439269 Chr.13: 26254806-26254806 on Build GRCh38
rs368416373 Chr.13: 26254829-26254829 on Build GRCh38
rs147657513 Chr.13: 26254737-26254737 on Build GRCh38
rs376295592 Chr.13: 26254804-26254804 on Build GRCh38
rs761720263 Chr.13: 26254789-26254789 on Build GRCh38
rs149166336 Chr.13: 26254740-26254740 on Build GRCh38
rs775184250 Chr.13: 26254774-26254775 on Build GRCh38
rs972837726 Chr.13: 26254825-26254825 on Build GRCh38
rs370721505 Chr.13: 26254816-26254816 on Build GRCh38
rs759556769 Chr.13: 26254768-26254769 on Build GRCh38
rs771706268 Chr.13: 26254770-26254771 on Build GRCh38
rs149938925 Chr.13: 26254796-26254796 on Build GRCh38
rs774087150 Chr.13: 26254766-26254767 on Build GRCh38
rs763947415 Chr.13: 26254778-26254778 on Build GRCh38
rs547006313 Chr.13: 26254792-26254792 on Build GRCh38
rs567350928 Chr.13: 26254807-26254807 on Build GRCh38
rs746009763 Chr.13: 26254802-26254802 on Build GRCh38
rs762864382 Chr.13: 26254755-26254755 on Build GRCh38
rs191644469 Chr.13: 26254779-26254779 on Build GRCh38

View Full Product Details