Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47232863-47233021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB2
Entrez Gene ID
1643
Gene Name
damage specific DNA binding protein 2
Gene Aliases
DDBB, UV-DDB2, XPE
UniGene
Hs.700338
Gene Chromosome Location
Chr.11: 47214942-47239218 on Build GRCh38
Transcripts
NM_000107.2 NM_001300734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363556 c.521C>T p.A174V Chr.11: 47232878-47232878 on Build GRCh38
927366 c.575G>A p.R192Q Chr.11: 47232932-47232932 on Build GRCh38
299932 c.537A>T p.G179G Chr.11: 47232894-47232894 on Build GRCh38
7122595 c.527C>T p.S176L Chr.11: 47232884-47232884 on Build GRCh38
687994 c.575G>T p.R192L Chr.11: 47232932-47232932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751753400 Chr.11: 47232942-47232942 on Build GRCh38
rs768530307 Chr.11: 47232919-47232919 on Build GRCh38
rs763313431 Chr.11: 47232932-47232932 on Build GRCh38
rs199822504 Chr.11: 47232931-47232931 on Build GRCh38
rs375488493 Chr.11: 47232968-47232968 on Build GRCh38
rs941873927 Chr.11: 47232981-47232981 on Build GRCh38
rs760106847 Chr.11: 47232944-47232944 on Build GRCh38
rs368460981 Chr.11: 47232996-47232996 on Build GRCh38
rs200406558 Chr.11: 47232934-47232934 on Build GRCh38
rs778251026 Chr.11: 47232987-47232987 on Build GRCh38
rs56042554 Chr.11: 47232979-47232979 on Build GRCh38
rs767854150 Chr.11: 47232953-47232953 on Build GRCh38
rs760322280 Chr.11: 47232890-47232890 on Build GRCh38
rs977177948 Chr.11: 47232908-47232908 on Build GRCh38
rs775515178 Chr.11: 47232886-47232886 on Build GRCh38
rs986048674 Chr.11: 47232978-47232978 on Build GRCh38
rs756970973 Chr.11: 47233001-47233001 on Build GRCh38

View Full Product Details