Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 22624596-22624754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCF
Entrez Gene ID
2188
Gene Name
Fanconi anemia complementation group F
Gene Aliases
FAF
UniGene
Hs.632151
Gene Chromosome Location
Chr.11: 22622533-22625841 on Build GRCh38
Transcripts
NM_022725.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1297926 c.1118C>T p.S373F Chr.11: 22624693-22624693 on Build GRCh38
4521389 c.1109G>A p.G370D Chr.11: 22624702-22624702 on Build GRCh38
1353252 c.1102A>G p.S368G Chr.11: 22624709-22624709 on Build GRCh38
4998387 c.1069G>T p.G357C Chr.11: 22624742-22624742 on Build GRCh38
5003954 c.1112T>G p.L371R Chr.11: 22624699-22624699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201285915 Chr.11: 22624724-22624724 on Build GRCh38
rs770063791 Chr.11: 22624655-22624655 on Build GRCh38
rs200505409 Chr.11: 22624670-22624670 on Build GRCh38
rs1029409487 Chr.11: 22624668-22624668 on Build GRCh38
rs764085506 Chr.11: 22624701-22624701 on Build GRCh38
rs756216888 Chr.11: 22624642-22624642 on Build GRCh38
rs748482579 Chr.11: 22624649-22624649 on Build GRCh38
rs781690040 Chr.11: 22624646-22624646 on Build GRCh38
rs372311535 Chr.11: 22624628-22624628 on Build GRCh38
rs770964562 Chr.11: 22624679-22624679 on Build GRCh38
rs763112358 Chr.11: 22624677-22624677 on Build GRCh38
rs544124149 Chr.11: 22624682-22624682 on Build GRCh38
rs753723379 Chr.11: 22624705-22624705 on Build GRCh38
rs760590516 Chr.11: 22624697-22624697 on Build GRCh38
rs761621055 Chr.11: 22624707-22624707 on Build GRCh38

View Full Product Details