Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40608762-40608894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KNL1
Entrez Gene ID
57082
Gene Name
kinetochore scaffold 1
Gene Aliases
AF15Q14, CASC5, CT29, D40, MCPH4, PPP1R55, Spc7, hKNL-1, hSpc105
UniGene
Hs.181855
Gene Chromosome Location
Chr.15: 40594012-40664342 on Build GRCh38
Transcripts
NM_144508.4 NM_170589.4 XM_017022432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5656460 c.177C>T p.V59V Chr.15: 40608888-40608888 on Build GRCh38
961229 c.169C>T p.R57C Chr.15: 40608880-40608880 on Build GRCh38
7447926 c.179G>A p.S60N Chr.15: 40608890-40608890 on Build GRCh38
3500914 c.141C>T p.S47S Chr.15: 40608852-40608852 on Build GRCh38
961231 c.172C>T p.R58* Chr.15: 40608883-40608883 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs891486869 Chr.15: 40608795-40608795 on Build GRCh38
rs754076742 Chr.15: 40608871-40608871 on Build GRCh38
rs1008508953 Chr.15: 40608822-40608822 on Build GRCh38
rs374869131 Chr.15: 40608832-40608832 on Build GRCh38
rs763938698 Chr.15: 40608855-40608855 on Build GRCh38
rs759490728 Chr.15: 40608826-40608826 on Build GRCh38
rs775359976 Chr.15: 40608854-40608854 on Build GRCh38
rs780341054 Chr.15: 40608873-40608873 on Build GRCh38
rs758641584 Chr.15: 40608870-40608870 on Build GRCh38
rs959944691 Chr.15: 40608802-40608802 on Build GRCh38
rs139813206 Chr.15: 40608793-40608793 on Build GRCh38
rs769524738 Chr.15: 40608868-40608870 on Build GRCh38
rs767317903 Chr.15: 40608843-40608843 on Build GRCh38
rs747727744 Chr.15: 40608808-40608809 on Build GRCh38

View Full Product Details