Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 187725505-187725638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL6
Entrez Gene ID
604
Gene Name
B-cell CLL/lymphoma 6
Gene Aliases
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
UniGene
Hs.478588
Gene Chromosome Location
Chr.3: 187721377-187745725 on Build GRCh38
Transcripts
NM_001130845.1 NM_001134738.1 NM_001706.4 XM_005247694.3 XM_011513062.2 NR_034062.1

Gene Symbol
LOC100131635
Entrez Gene ID
100131635
Gene Name
hCG1645011-like
Gene Aliases
-
UniGene
Hs.434448
Gene Chromosome Location
Chr.3: 187702366-187732415 on Build GRCh38
Transcripts
NM_001130845.1 NM_001134738.1 NM_001706.4 XM_005247694.3 XM_011513062.2 NR_034062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1042447 c.1709-1G>A p.? Chr.3: 187725630-187725630 on Build GRCh38
6610727 c.1716A>G p.K572K Chr.3: 187725622-187725622 on Build GRCh38
1421597 c.1812C>T p.C604C Chr.3: 187725526-187725526 on Build GRCh38
6999804 c.1822G>A p.G608R Chr.3: 187725516-187725516 on Build GRCh38
5817657 c.1812C>G p.C604W Chr.3: 187725526-187725526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528353546 Chr.3: 187725574-187725574 on Build GRCh38
rs1032264970 Chr.3: 187725580-187725580 on Build GRCh38
rs922428901 Chr.3: 187725550-187725550 on Build GRCh38
rs753064923 Chr.3: 187725604-187725604 on Build GRCh38
rs996848361 Chr.3: 187725585-187725585 on Build GRCh38
rs61732778 Chr.3: 187725526-187725526 on Build GRCh38
rs760062751 Chr.3: 187725565-187725565 on Build GRCh38
rs767829359 Chr.3: 187725583-187725583 on Build GRCh38
rs752044631 Chr.3: 187725558-187725558 on Build GRCh38

View Full Product Details