Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 8055836-8056000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA3
Entrez Gene ID
2625
Gene Name
GATA binding protein 3
Gene Aliases
HDR, HDRS
UniGene
Hs.524134
Gene Chromosome Location
Chr.10: 8045421-8075201 on Build GRCh38
Transcripts
NM_001002295.1 NM_002051.2 XM_005252442.2 XM_005252443.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5703729 c.241+2T>C p.? Chr.10: 8055898-8055898 on Build GRCh38
4771385 c.200C>G p.S67W Chr.10: 8055855-8055855 on Build GRCh38
3738746 c.230C>T p.P77L Chr.10: 8055885-8055885 on Build GRCh38
5703727 c.241+1G>C p.? Chr.10: 8055897-8055897 on Build GRCh38
5703728 c.241+1G>T p.? Chr.10: 8055897-8055897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755756354 Chr.10: 8055881-8055881 on Build GRCh38
rs757596536 Chr.10: 8055929-8055929 on Build GRCh38
rs1037111200 Chr.10: 8055926-8055926 on Build GRCh38
rs753449980 Chr.10: 8055905-8055905 on Build GRCh38
rs762338199 Chr.10: 8055978-8055978 on Build GRCh38
rs1004765920 Chr.10: 8055888-8055888 on Build GRCh38
rs778031846 Chr.10: 8055917-8055917 on Build GRCh38
rs997332766 Chr.10: 8055946-8055946 on Build GRCh38
rs774154154 Chr.10: 8055860-8055860 on Build GRCh38
rs767609253 Chr.10: 8055868-8055868 on Build GRCh38
rs1030189256 Chr.10: 8055971-8055971 on Build GRCh38
rs561659655 Chr.10: 8055977-8055977 on Build GRCh38
rs759839816 Chr.10: 8055896-8055896 on Build GRCh38
rs369275197 Chr.10: 8055925-8055925 on Build GRCh38
rs560531559 Chr.10: 8055871-8055871 on Build GRCh38
rs932686919 Chr.10: 8055861-8055861 on Build GRCh38
rs754594895 Chr.10: 8055908-8055908 on Build GRCh38
rs745825837 Chr.10: 8055937-8055938 on Build GRCh38
rs376505863 Chr.10: 8055904-8055904 on Build GRCh38
rs761676499 Chr.10: 8055864-8055864 on Build GRCh38

View Full Product Details