Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94781806-94781945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C19
Entrez Gene ID
1557
Gene Name
cytochrome P450 family 2 subfamily C member 19
Gene Aliases
CPCJ, CYP2C, CYPIIC17, CYPIIC19, P450C2C, P450IIC19
UniGene
Hs.282409
Gene Chromosome Location
Chr.10: 94762681-94853205 on Build GRCh38
Transcripts
NM_000769.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5915922 c.727G>A p.D243N Chr.10: 94781905-94781905 on Build GRCh38
3441954 c.658C>T p.P220S Chr.10: 94781836-94781836 on Build GRCh38
686015 c.757G>A p.E253K Chr.10: 94781935-94781935 on Build GRCh38
1970806 c.762G>A p.S254S Chr.10: 94781940-94781940 on Build GRCh38
686016 c.688C>T p.H230Y Chr.10: 94781866-94781866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763972230 Chr.10: 94781906-94781906 on Build GRCh38
rs267602635 Chr.10: 94781844-94781844 on Build GRCh38
rs879130837 Chr.10: 94781858-94781859 on Build GRCh38
rs767307799 Chr.10: 94781891-94781891 on Build GRCh38
rs963088247 Chr.10: 94781846-94781846 on Build GRCh38
rs574458036 Chr.10: 94781916-94781916 on Build GRCh38
rs771243716 Chr.10: 94781886-94781886 on Build GRCh38
rs745394523 Chr.10: 94781841-94781841 on Build GRCh38
rs759690171 Chr.10: 94781890-94781890 on Build GRCh38
rs775544863 Chr.10: 94781899-94781899 on Build GRCh38
rs760734417 Chr.10: 94781904-94781904 on Build GRCh38
rs768974528 Chr.10: 94781849-94781849 on Build GRCh38
rs954545346 Chr.10: 94781877-94781877 on Build GRCh38
rs72558185 Chr.10: 94781899-94781900 on Build GRCh38
rs774238831 Chr.10: 94781889-94781889 on Build GRCh38
rs6413438 Chr.10: 94781858-94781858 on Build GRCh38
rs781265742 Chr.10: 94781914-94781914 on Build GRCh38
rs866297301 Chr.10: 94781855-94781855 on Build GRCh38
rs4244285 Chr.10: 94781859-94781859 on Build GRCh38
rs749507932 Chr.10: 94781885-94781885 on Build GRCh38
rs770126483 Chr.10: 94781871-94781871 on Build GRCh38
rs566311971 Chr.10: 94781838-94781838 on Build GRCh38
rs200362177 Chr.10: 94781854-94781854 on Build GRCh38

View Full Product Details