Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 52923678-52923808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCC
Entrez Gene ID
1630
Gene Name
DCC netrin 1 receptor
Gene Aliases
CRC18, CRCR1, IGDCC1, MRMV1, NTN1R1
UniGene
Hs.162025
Gene Chromosome Location
Chr.18: 52340172-53535903 on Build GRCh38
Transcripts
NM_005215.3 XM_011525843.1 XM_017025568.1 XM_017025569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5758232 c.767A>T p.D256V Chr.18: 52923776-52923776 on Build GRCh38
391576 c.762delA p.D256fs*45 Chr.18: 52923771-52923771 on Build GRCh38
3526319 c.727C>T p.L243L Chr.18: 52923736-52923736 on Build GRCh38
4455371 c.747A>G p.V249V Chr.18: 52923756-52923756 on Build GRCh38
4580447 c.281C>T p.P94L Chr.18: 52923746-52923746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771073523 Chr.18: 52923764-52923764 on Build GRCh38
rs755869531 Chr.18: 52923752-52923752 on Build GRCh38
rs201234438 Chr.18: 52923718-52923718 on Build GRCh38
rs747189622 Chr.18: 52923758-52923758 on Build GRCh38
rs759718173 Chr.18: 52923775-52923775 on Build GRCh38
rs745379903 Chr.18: 52923726-52923726 on Build GRCh38
rs751210422 Chr.18: 52923710-52923710 on Build GRCh38
rs376124257 Chr.18: 52923770-52923770 on Build GRCh38
rs986247204 Chr.18: 52923779-52923779 on Build GRCh38
rs764081462 Chr.18: 52923755-52923755 on Build GRCh38
rs765447173 Chr.18: 52923777-52923777 on Build GRCh38
rs776117215 Chr.18: 52923780-52923780 on Build GRCh38
rs769667349 Chr.18: 52923728-52923728 on Build GRCh38
rs981733248 Chr.18: 52923784-52923784 on Build GRCh38
rs779938904 Chr.18: 52923738-52923738 on Build GRCh38
rs146026074 Chr.18: 52923715-52923715 on Build GRCh38
rs139558399 Chr.18: 52923753-52923753 on Build GRCh38
rs537417676 Chr.18: 52923781-52923781 on Build GRCh38

View Full Product Details