Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 112224715-112224868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD3
Entrez Gene ID
114788
Gene Name
CUB and Sushi multiple domains 3
Gene Aliases
-
UniGene
Hs.91381
Gene Chromosome Location
Chr.8: 112222928-113437499 on Build GRCh38
Transcripts
NM_052900.2 NM_198123.1 NM_198124.1 XM_011516815.2 XM_011516816.2 XM_017013008.1 XM_017013009.1 XM_017013010.1 XM_017013011.1 XM_017013012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3770025 c.10987G>T p.V3663F Chr.8: 112224788-112224788 on Build GRCh38
5184404 c.11055C>T p.N3685N Chr.8: 112224840-112224840 on Build GRCh38
6111933 c.10935C>A p.N3645K Chr.8: 112224840-112224840 on Build GRCh38
5710816 c.10951G>A p.G3651R Chr.8: 112224824-112224824 on Build GRCh38
3884786 c.10936G>A p.A3646T Chr.8: 112224839-112224839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747508555 Chr.8: 112224811-112224811 on Build GRCh38
rs767515522 Chr.8: 112224761-112224761 on Build GRCh38
rs879134387 Chr.8: 112224834-112224834 on Build GRCh38
rs4876458 Chr.8: 112224816-112224816 on Build GRCh38
rs758783402 Chr.8: 112224803-112224803 on Build GRCh38
rs767462115 Chr.8: 112224838-112224838 on Build GRCh38
rs755097432 Chr.8: 112224789-112224789 on Build GRCh38
rs753793600 Chr.8: 112224841-112224841 on Build GRCh38
rs144647532 Chr.8: 112224796-112224796 on Build GRCh38
rs768386869 Chr.8: 112224817-112224817 on Build GRCh38
rs748470778 Chr.8: 112224801-112224801 on Build GRCh38
rs760587971 Chr.8: 112224840-112224840 on Build GRCh38
rs375360016 Chr.8: 112224749-112224749 on Build GRCh38
rs778227550 Chr.8: 112224809-112224809 on Build GRCh38
rs150947415 Chr.8: 112224770-112224770 on Build GRCh38
rs754078265 Chr.8: 112224780-112224780 on Build GRCh38
rs1040825656 Chr.8: 112224762-112224762 on Build GRCh38
rs369702665 Chr.8: 112224750-112224750 on Build GRCh38
rs769337797 Chr.8: 112224812-112224812 on Build GRCh38
rs766437541 Chr.8: 112224772-112224772 on Build GRCh38
rs891524968 Chr.8: 112224798-112224798 on Build GRCh38
rs940441521 Chr.8: 112224804-112224804 on Build GRCh38
rs773198324 Chr.8: 112224839-112224839 on Build GRCh38
rs774172040 Chr.8: 112224818-112224818 on Build GRCh38
rs373200529 Chr.8: 112224815-112224815 on Build GRCh38
rs763855862 Chr.8: 112224746-112224746 on Build GRCh38

View Full Product Details