Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42127762-42127906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2D6
Entrez Gene ID
1565
Gene Name
cytochrome P450 family 2 subfamily D member 6
Gene Aliases
CPD6, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, P450-DB1, P450C2D, P450DB1
UniGene
Hs.648256
Gene Chromosome Location
Chr.22: 42125531-42130881 on Build GRCh38
Transcripts
NM_000106.5 NM_001025161.2 XM_011529966.2 XM_011529968.2 XM_011529970.2 XM_011529972.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6639222 c.933G>A p.S311S Chr.22: 42127894-42127894 on Build GRCh38
6639224 c.926C>T p.T309I Chr.22: 42127901-42127901 on Build GRCh38
4104553 c.799C>A p.L267I Chr.22: 42127875-42127875 on Build GRCh38
6381676 c.796G>T p.G266C Chr.22: 42127878-42127878 on Build GRCh38
5859498 c.921G>A p.M307I Chr.22: 42127906-42127906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148250267 Chr.22: 42127833-42127833 on Build GRCh38
rs113420533 Chr.22: 42127839-42127839 on Build GRCh38
rs777955329 Chr.22: 42127846-42127846 on Build GRCh38
rs28371725 Chr.22: 42127803-42127803 on Build GRCh38
rs369999094 Chr.22: 42127810-42127810 on Build GRCh38
rs141289473 Chr.22: 42127869-42127869 on Build GRCh38
rs372910785 Chr.22: 42127851-42127851 on Build GRCh38
rs28371724 Chr.22: 42127855-42127855 on Build GRCh38
rs749933540 Chr.22: 42127793-42127793 on Build GRCh38
rs72549349 Chr.22: 42127841-42127841 on Build GRCh38
rs373844237 Chr.22: 42127799-42127799 on Build GRCh38
rs377167058 Chr.22: 42127888-42127888 on Build GRCh38
rs780137937 Chr.22: 42127796-42127796 on Build GRCh38
rs369176838 Chr.22: 42127849-42127849 on Build GRCh38
rs747917111 Chr.22: 42127804-42127804 on Build GRCh38
rs5030867 Chr.22: 42127856-42127856 on Build GRCh38
rs140513104 Chr.22: 42127853-42127853 on Build GRCh38
rs746324085 Chr.22: 42127813-42127813 on Build GRCh38
rs754895235 Chr.22: 42127835-42127835 on Build GRCh38
rs730882170 Chr.22: 42127846-42127864 on Build GRCh38
rs778008354 Chr.22: 42127808-42127808 on Build GRCh38
rs143276168 Chr.22: 42127811-42127811 on Build GRCh38
rs554540676 Chr.22: 42127822-42127822 on Build GRCh38
rs747397644 Chr.22: 42127861-42127861 on Build GRCh38
rs149528658 Chr.22: 42127870-42127870 on Build GRCh38
rs773066646 Chr.22: 42127828-42127828 on Build GRCh38
rs764953443 Chr.22: 42127827-42127827 on Build GRCh38
rs775904145 Chr.22: 42127857-42127857 on Build GRCh38
rs772490933 Chr.22: 42127818-42127818 on Build GRCh38
rs746736981 Chr.22: 42127821-42127822 on Build GRCh38
rs150216909 Chr.22: 42127842-42127842 on Build GRCh38
rs773889591 Chr.22: 42127874-42127874 on Build GRCh38
rs767401994 Chr.22: 42127837-42127837 on Build GRCh38
rs201895960 Chr.22: 42127831-42127831 on Build GRCh38
rs550835698 Chr.22: 42127830-42127831 on Build GRCh38
rs777148766 Chr.22: 42127867-42127867 on Build GRCh38
rs192090178 Chr.22: 42127807-42127807 on Build GRCh38
rs143145507 Chr.22: 42127889-42127889 on Build GRCh38
rs200234159 Chr.22: 42127865-42127865 on Build GRCh38
rs760996241 Chr.22: 42127826-42127826 on Build GRCh38
rs965370012 Chr.22: 42127878-42127878 on Build GRCh38
rs765006570 Chr.22: 42127792-42127792 on Build GRCh38
rs187334823 Chr.22: 42127786-42127786 on Build GRCh38
rs953106263 Chr.22: 42127850-42127850 on Build GRCh38
rs1014423327 Chr.22: 42127821-42127821 on Build GRCh38
rs569426051 Chr.22: 42127790-42127790 on Build GRCh38
rs375586652 Chr.22: 42127876-42127876 on Build GRCh38
rs536709258 Chr.22: 42127791-42127791 on Build GRCh38
rs751061716 Chr.22: 42127832-42127832 on Build GRCh38
rs79292917 Chr.22: 42127852-42127852 on Build GRCh38
rs281860295 Chr.22: 42127864-42127882 on Build GRCh38
rs767483427 Chr.22: 42127877-42127877 on Build GRCh38
rs879550940 Chr.22: 42127855-42127856 on Build GRCh38

View Full Product Details