Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100807327-100807472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB4
Entrez Gene ID
2050
Gene Name
EPH receptor B4
Gene Aliases
HTK, MYK1, TYRO11
UniGene
Hs.437008
Gene Chromosome Location
Chr.7: 100802565-100827521 on Build GRCh38
Transcripts
NM_004444.4 XM_017011816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5574606 c.2300A>G p.E767G Chr.7: 100807399-100807399 on Build GRCh38
6202136 c.2316C>T p.P772P Chr.7: 100807383-100807383 on Build GRCh38
743650 c.2280C>T p.G760G Chr.7: 100807419-100807419 on Build GRCh38
3080851 c.2324C>T p.T775M Chr.7: 100807375-100807375 on Build GRCh38
6655263 c.2311G>A p.D771N Chr.7: 100807388-100807388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146460992 Chr.7: 100807356-100807356 on Build GRCh38
rs143966632 Chr.7: 100807377-100807377 on Build GRCh38
rs368484374 Chr.7: 100807442-100807442 on Build GRCh38
rs750453997 Chr.7: 100807408-100807408 on Build GRCh38
rs200376692 Chr.7: 100807447-100807447 on Build GRCh38
rs764351124 Chr.7: 100807390-100807390 on Build GRCh38
rs777063679 Chr.7: 100807391-100807391 on Build GRCh38
rs147302485 Chr.7: 100807389-100807389 on Build GRCh38
rs750554256 Chr.7: 100807349-100807349 on Build GRCh38
rs576306239 Chr.7: 100807443-100807443 on Build GRCh38
rs371509217 Chr.7: 100807407-100807407 on Build GRCh38
rs762159059 Chr.7: 100807392-100807392 on Build GRCh38
rs373776011 Chr.7: 100807419-100807419 on Build GRCh38
rs765763445 Chr.7: 100807406-100807406 on Build GRCh38
rs554530916 Chr.7: 100807431-100807431 on Build GRCh38
rs374120458 Chr.7: 100807368-100807368 on Build GRCh38
rs772783851 Chr.7: 100807361-100807361 on Build GRCh38
rs35809853 Chr.7: 100807371-100807372 on Build GRCh38
rs528672824 Chr.7: 100807422-100807422 on Build GRCh38
rs984719753 Chr.7: 100807380-100807380 on Build GRCh38
rs367628554 Chr.7: 100807375-100807375 on Build GRCh38
rs747983713 Chr.7: 100807446-100807446 on Build GRCh38

View Full Product Details