Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94762812-94762969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C19
Entrez Gene ID
1557
Gene Name
cytochrome P450 family 2 subfamily C member 19
Gene Aliases
CPCJ, CYP2C, CYPIIC17, CYPIIC19, P450C2C, P450IIC19
UniGene
Hs.282409
Gene Chromosome Location
Chr.10: 94762681-94853205 on Build GRCh38
Transcripts
NM_000769.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1603878 c.168+7A>G p.? Chr.10: 94762880-94762880 on Build GRCh38
7231805 c.112G>C p.V38L Chr.10: 94762817-94762817 on Build GRCh38
3808063 c.147T>A p.D49E Chr.10: 94762852-94762852 on Build GRCh38
6639196 c.125T>C p.I42T Chr.10: 94762830-94762830 on Build GRCh38
4470437 c.165C>T p.T55T Chr.10: 94762870-94762870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910785959 Chr.10: 94762929-94762929 on Build GRCh38
rs17882201 Chr.10: 94762888-94762888 on Build GRCh38
rs777031765 Chr.10: 94762881-94762881 on Build GRCh38
rs755882715 Chr.10: 94762842-94762842 on Build GRCh38
rs777673347 Chr.10: 94762852-94762852 on Build GRCh38
rs372709413 Chr.10: 94762910-94762910 on Build GRCh38
rs766303560 Chr.10: 94762886-94762888 on Build GRCh38
rs57079188 Chr.10: 94762901-94762901 on Build GRCh38
rs769135473 Chr.10: 94762880-94762880 on Build GRCh38
rs17881883 Chr.10: 94762893-94762893 on Build GRCh38
rs748915758 Chr.10: 94762853-94762853 on Build GRCh38
rs931824204 Chr.10: 94762898-94762898 on Build GRCh38
rs745382759 Chr.10: 94762873-94762873 on Build GRCh38
rs766647991 Chr.10: 94762902-94762902 on Build GRCh38
rs944797319 Chr.10: 94762914-94762914 on Build GRCh38
rs530292726 Chr.10: 94762922-94762922 on Build GRCh38
rs943657018 Chr.10: 94762939-94762939 on Build GRCh38
rs572853437 Chr.10: 94762869-94762869 on Build GRCh38
rs755017877 Chr.10: 94762923-94762923 on Build GRCh38
rs765501102 Chr.10: 94762889-94762889 on Build GRCh38

View Full Product Details