Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77794549-77794705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAQ
Entrez Gene ID
2776
Gene Name
G protein subunit alpha q
Gene Aliases
CMC1, G-ALPHA-q, GAQ, SWS
UniGene
Hs.269782
Gene Chromosome Location
Chr.9: 77716274-78031449 on Build GRCh38
Transcripts
NM_002072.4 XM_017014628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6580214 c.647G>A p.W216* Chr.9: 77794551-77794551 on Build GRCh38
6342247 c.645_645delA p.K215fs*15 Chr.9: 77794553-77794553 on Build GRCh38
1701256 c.632C>T p.S211L Chr.9: 77794566-77794566 on Build GRCh38
1110326 c.612C>T p.V204V Chr.9: 77794586-77794586 on Build GRCh38
6342244 c.620G>A p.G207E Chr.9: 77794578-77794578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771098223 Chr.9: 77794634-77794634 on Build GRCh38
rs777924414 Chr.9: 77794625-77794625 on Build GRCh38
rs1028716428 Chr.9: 77794668-77794668 on Build GRCh38
rs775333366 Chr.9: 77794636-77794636 on Build GRCh38
rs979405827 Chr.9: 77794649-77794649 on Build GRCh38
rs769875049 Chr.9: 77794615-77794615 on Build GRCh38
rs781615478 Chr.9: 77794612-77794612 on Build GRCh38
rs371519995 Chr.9: 77794577-77794577 on Build GRCh38
rs749544720 Chr.9: 77794632-77794632 on Build GRCh38
rs755497004 Chr.9: 77794611-77794611 on Build GRCh38
rs780431354 Chr.9: 77794586-77794586 on Build GRCh38
rs754010787 Chr.9: 77794607-77794607 on Build GRCh38
rs748158046 Chr.9: 77794613-77794613 on Build GRCh38

View Full Product Details