Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47375209-47375366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMPK1
Entrez Gene ID
51727
Gene Name
cytidine/uridine monophosphate kinase 1
Gene Aliases
CK, CMK, CMPK, UMK, UMP-CMPK, UMPK
UniGene
Hs.731647
Gene Chromosome Location
Chr.1: 47333797-47378839 on Build GRCh38
Transcripts
NM_001136140.1 NM_016308.2 NR_046394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
398125 c.572C>A p.T191K Chr.1: 47375220-47375220 on Build GRCh38
6382028 c.628T>C p.S210P Chr.1: 47375276-47375276 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766232563 Chr.1: 47375270-47375270 on Build GRCh38
rs752778206 Chr.1: 47375326-47375326 on Build GRCh38
rs759415022 Chr.1: 47375308-47375308 on Build GRCh38
rs760432890 Chr.1: 47375249-47375249 on Build GRCh38
rs915362521 Chr.1: 47375235-47375235 on Build GRCh38
rs1043079088 Chr.1: 47375256-47375256 on Build GRCh38
rs1057001101 Chr.1: 47375321-47375321 on Build GRCh38
rs905419751 Chr.1: 47375298-47375299 on Build GRCh38
rs776538314 Chr.1: 47375278-47375278 on Build GRCh38
rs745668983 Chr.1: 47375323-47375324 on Build GRCh38
rs769524257 Chr.1: 47375285-47375285 on Build GRCh38
rs765087140 Chr.1: 47375313-47375313 on Build GRCh38

View Full Product Details