Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33318977-33319151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAXX
Entrez Gene ID
1616
Gene Name
death domain associated protein
Gene Aliases
BING2, DAP6, EAP1
UniGene
Hs.336916
Gene Chromosome Location
Chr.6: 33318558-33323016 on Build GRCh38
Transcripts
NM_001141969.1 NM_001141970.1 NM_001254717.1 NM_001350.4 XM_005248860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6969147 c.2045C>T p.S682F Chr.6: 33319115-33319115 on Build GRCh38
85803 c.2120_2120delC p.Q708fs*34 Chr.6: 33319040-33319040 on Build GRCh38
4676233 c.2025C>A p.S675S Chr.6: 33319135-33319135 on Build GRCh38
3625706 c.2019G>A p.L673L Chr.6: 33319141-33319141 on Build GRCh38
5019541 c.2112C>T p.A704A Chr.6: 33319048-33319048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140525877 Chr.6: 33319129-33319129 on Build GRCh38
rs143737807 Chr.6: 33319046-33319046 on Build GRCh38
rs769706471 Chr.6: 33319049-33319049 on Build GRCh38
rs768007918 Chr.6: 33319119-33319119 on Build GRCh38
rs377663648 Chr.6: 33319115-33319115 on Build GRCh38
rs369424162 Chr.6: 33319014-33319014 on Build GRCh38
rs150309352 Chr.6: 33319042-33319042 on Build GRCh38
rs555856847 Chr.6: 33319047-33319047 on Build GRCh38
rs755784520 Chr.6: 33319031-33319031 on Build GRCh38
rs915933778 Chr.6: 33319040-33319040 on Build GRCh38
rs752093917 Chr.6: 33319013-33319013 on Build GRCh38
rs755659352 Chr.6: 33319126-33319126 on Build GRCh38
rs1026271281 Chr.6: 33319028-33319028 on Build GRCh38
rs753556048 Chr.6: 33319130-33319130 on Build GRCh38
rs376435137 Chr.6: 33319000-33319000 on Build GRCh38
rs137897838 Chr.6: 33319043-33319043 on Build GRCh38
rs780728486 Chr.6: 33319019-33319019 on Build GRCh38
rs1801849 Chr.6: 33319021-33319021 on Build GRCh38
rs762346785 Chr.6: 33319109-33319109 on Build GRCh38
rs749955449 Chr.6: 33319125-33319125 on Build GRCh38
rs751898991 Chr.6: 33319106-33319106 on Build GRCh38
rs371458373 Chr.6: 33319068-33319068 on Build GRCh38
rs951934351 Chr.6: 33319033-33319033 on Build GRCh38
rs200069239 Chr.6: 33319066-33319066 on Build GRCh38
rs749903797 Chr.6: 33319022-33319022 on Build GRCh38
rs11873 Chr.6: 33319048-33319048 on Build GRCh38
rs775787032 Chr.6: 33319053-33319053 on Build GRCh38
rs778805230 Chr.6: 33319045-33319045 on Build GRCh38
rs924778264 Chr.6: 33319075-33319075 on Build GRCh38
rs763046918 Chr.6: 33319055-33319055 on Build GRCh38

View Full Product Details