Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 89340902-89341033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA3
Entrez Gene ID
2042
Gene Name
EPH receptor A3
Gene Aliases
EK4, ETK, ETK1, HEK, HEK4, TYRO4
UniGene
Hs.123642
Gene Chromosome Location
Chr.3: 89107524-89482134 on Build GRCh38
Transcripts
NM_005233.5 NM_182644.2 XM_005264715.2 XM_005264716.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6987368 c.833A>G p.Y278C Chr.3: 89340934-89340934 on Build GRCh38
6165564 c.834C>A p.Y278* Chr.3: 89340935-89340935 on Build GRCh38
731475 c.834C>T p.Y278Y Chr.3: 89340935-89340935 on Build GRCh38
6970429 c.906G>A p.M302I Chr.3: 89341007-89341007 on Build GRCh38
6273666 c.899G>T p.G300V Chr.3: 89341000-89341000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182691254 Chr.3: 89340942-89340942 on Build GRCh38
rs751588436 Chr.3: 89340935-89340935 on Build GRCh38
rs758077460 Chr.3: 89340958-89340958 on Build GRCh38
rs781383365 Chr.3: 89340947-89340947 on Build GRCh38
rs1056467539 Chr.3: 89340929-89340929 on Build GRCh38
rs764312912 Chr.3: 89340928-89340928 on Build GRCh38
rs755010537 Chr.3: 89340948-89340948 on Build GRCh38
rs948106621 Chr.3: 89340957-89340957 on Build GRCh38
rs747973326 Chr.3: 89340955-89340955 on Build GRCh38
rs896512954 Chr.3: 89340940-89340940 on Build GRCh38
rs142109561 Chr.3: 89340993-89340993 on Build GRCh38
rs200484511 Chr.3: 89340951-89340951 on Build GRCh38

View Full Product Details