Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 68638927-68639091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRB3
Entrez Gene ID
577
Gene Name
adhesion G protein-coupled receptor B3
Gene Aliases
BAI3
UniGene
Hs.13261
Gene Chromosome Location
Chr.6: 68635282-69389511 on Build GRCh38
Transcripts
NM_001704.2 XM_005248752.2 XM_006715534.3 XM_011536010.2 XM_011536011.2 XM_011536012.2 XM_017011140.1 XM_017011142.1 XM_017011143.1 XM_017011144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5405722 c.416C>T p.P139L Chr.6: 68639091-68639091 on Build GRCh38
5357731 c.280T>C p.S94P Chr.6: 68638955-68638955 on Build GRCh38
309423 c.396A>T p.R132R Chr.6: 68639071-68639071 on Build GRCh38
6450082 c.392G>A p.R131H Chr.6: 68639067-68639067 on Build GRCh38
3160726 c.329A>C p.Q110P Chr.6: 68639004-68639004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377083198 Chr.6: 68638966-68638966 on Build GRCh38
rs759795908 Chr.6: 68638972-68638972 on Build GRCh38
rs767760754 Chr.6: 68638973-68638973 on Build GRCh38
rs754603670 Chr.6: 68639029-68639029 on Build GRCh38
rs756724848 Chr.6: 68638986-68638986 on Build GRCh38
rs746699209 Chr.6: 68639020-68639020 on Build GRCh38
rs553411317 Chr.6: 68639016-68639016 on Build GRCh38
rs267601100 Chr.6: 68638956-68638956 on Build GRCh38
rs770693543 Chr.6: 68639064-68639064 on Build GRCh38
rs1014728193 Chr.6: 68639050-68639050 on Build GRCh38
rs764728357 Chr.6: 68638987-68638987 on Build GRCh38
rs774854338 Chr.6: 68638960-68638960 on Build GRCh38
rs1031211547 Chr.6: 68638970-68638970 on Build GRCh38
rs752799769 Chr.6: 68638975-68638975 on Build GRCh38
rs770588704 Chr.6: 68638972-68638972 on Build GRCh38
rs749186524 Chr.6: 68639057-68639057 on Build GRCh38
rs573441734 Chr.6: 68639026-68639026 on Build GRCh38
rs190415498 Chr.6: 68639036-68639036 on Build GRCh38
rs12198940 Chr.6: 68639032-68639032 on Build GRCh38
rs769247421 Chr.6: 68639046-68639046 on Build GRCh38
rs773040433 Chr.6: 68639052-68639052 on Build GRCh38
rs754321363 Chr.6: 68638990-68638990 on Build GRCh38
rs895562853 Chr.6: 68638991-68638991 on Build GRCh38
rs780722343 Chr.6: 68639030-68639030 on Build GRCh38
rs757575126 Chr.6: 68639001-68639001 on Build GRCh38
rs773163654 Chr.6: 68638957-68638957 on Build GRCh38
rs774436786 Chr.6: 68638967-68638967 on Build GRCh38

View Full Product Details