Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215365405-215365579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FN1
Entrez Gene ID
2335
Gene Name
fibronectin 1
Gene Aliases
CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
UniGene
Hs.203717
Gene Chromosome Location
Chr.2: 215360440-215436167 on Build GRCh38
Transcripts
NM_001306129.1 NM_001306130.1 NM_001306131.1 NM_001306132.1 NM_002026.3 NM_212474.2 NM_212476.2 NM_212478.2 NM_212482.2 XM_005246397.1 XM_005246398.1 XM_005246399.1 XM_005246401.1 XM_005246402.1 XM_005246403.1 XM_005246404.1 XM_005246405.1 XM_005246406.1 XM_005246407.1 XM_005246408.1 XM_005246409.1 XM_005246410.1 XM_005246411.1 XM_005246412.1 XM_005246414.1 XM_005246416.1 XM_017003692.1 XM_017003693.1 XM_017003694.1 XM_017003695.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7144652 c.7126G>A p.E2376K Chr.2: 215365523-215365523 on Build GRCh38
1136598 c.7114A>G p.N2372D Chr.2: 215365535-215365535 on Build GRCh38
1591863 c.7101C>T p.C2367C Chr.2: 215365548-215365548 on Build GRCh38
7156835 c.6726G>A p.M2242I Chr.2: 215365557-215365557 on Build GRCh38
7156834 c.7092G>A p.M2364I Chr.2: 215365557-215365557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031354568 Chr.2: 215365461-215365461 on Build GRCh38
rs754861847 Chr.2: 215365532-215365532 on Build GRCh38
rs112327569 Chr.2: 215365502-215365502 on Build GRCh38
rs776878703 Chr.2: 215365466-215365466 on Build GRCh38
rs765551193 Chr.2: 215365488-215365488 on Build GRCh38
rs373954333 Chr.2: 215365494-215365494 on Build GRCh38
rs998579993 Chr.2: 215365445-215365445 on Build GRCh38
rs10498037 Chr.2: 215365493-215365493 on Build GRCh38
rs751589162 Chr.2: 215365501-215365501 on Build GRCh38
rs201635143 Chr.2: 215365545-215365545 on Build GRCh38
rs749068438 Chr.2: 215365531-215365531 on Build GRCh38
rs890752814 Chr.2: 215365439-215365439 on Build GRCh38
rs112323657 Chr.2: 215365504-215365504 on Build GRCh38
rs568449322 Chr.2: 215365533-215365533 on Build GRCh38
rs548828880 Chr.2: 215365554-215365554 on Build GRCh38
rs748159065 Chr.2: 215365544-215365544 on Build GRCh38
rs957105203 Chr.2: 215365481-215365481 on Build GRCh38
rs759900198 Chr.2: 215365469-215365469 on Build GRCh38
rs768302524 Chr.2: 215365518-215365518 on Build GRCh38

View Full Product Details