Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94762606-94762761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C19
Entrez Gene ID
1557
Gene Name
cytochrome P450 family 2 subfamily C member 19
Gene Aliases
CPCJ, CYP2C, CYPIIC17, CYPIIC19, P450C2C, P450IIC19
UniGene
Hs.282409
Gene Chromosome Location
Chr.10: 94762681-94853205 on Build GRCh38
Transcripts
NM_000769.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
686019 c.48C>T p.L16L Chr.10: 94762753-94762753 on Build GRCh38
1702795 c.35C>T p.S12L Chr.10: 94762740-94762740 on Build GRCh38
3709965 c.8C>T p.P3L Chr.10: 94762713-94762713 on Build GRCh38
1721782 c.7C>T p.P3S Chr.10: 94762712-94762712 on Build GRCh38
4746065 c.42G>T p.L14F Chr.10: 94762747-94762747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377381376 Chr.10: 94762709-94762709 on Build GRCh38
rs757485026 Chr.10: 94762728-94762728 on Build GRCh38
rs764485894 Chr.10: 94762724-94762724 on Build GRCh38
rs889254628 Chr.10: 94762703-94762703 on Build GRCh38
rs555087130 Chr.10: 94762700-94762700 on Build GRCh38
rs1036765817 Chr.10: 94762645-94762645 on Build GRCh38
rs750431855 Chr.10: 94762730-94762730 on Build GRCh38
rs28399504 Chr.10: 94762706-94762706 on Build GRCh38
rs768096659 Chr.10: 94762713-94762713 on Build GRCh38
rs200381600 Chr.10: 94762733-94762733 on Build GRCh38
rs566121294 Chr.10: 94762646-94762646 on Build GRCh38
rs1014878497 Chr.10: 94762643-94762643 on Build GRCh38
rs1009356509 Chr.10: 94762717-94762717 on Build GRCh38
rs991546782 Chr.10: 94762631-94762631 on Build GRCh38
rs933536968 Chr.10: 94762678-94762678 on Build GRCh38
rs141890453 Chr.10: 94762727-94762727 on Build GRCh38
rs367543001 Chr.10: 94762693-94762693 on Build GRCh38
rs746770214 Chr.10: 94762677-94762677 on Build GRCh38
rs779012951 Chr.10: 94762729-94762729 on Build GRCh38
rs367543003 Chr.10: 94762715-94762715 on Build GRCh38
rs779922341 Chr.10: 94762735-94762735 on Build GRCh38
rs761311559 Chr.10: 94762720-94762720 on Build GRCh38
rs768465675 Chr.10: 94762683-94762683 on Build GRCh38
rs771935348 Chr.10: 94762662-94762662 on Build GRCh38
rs17886301 Chr.10: 94762636-94762636 on Build GRCh38
rs779975527 Chr.10: 94762676-94762676 on Build GRCh38
rs1021000556 Chr.10: 94762734-94762734 on Build GRCh38
rs763763633 Chr.10: 94762644-94762644 on Build GRCh38
rs367543002 Chr.10: 94762712-94762712 on Build GRCh38
rs745989870 Chr.10: 94762657-94762657 on Build GRCh38
rs754219797 Chr.10: 94762725-94762725 on Build GRCh38

View Full Product Details