Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33319108-33319257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAXX
Entrez Gene ID
1616
Gene Name
death domain associated protein
Gene Aliases
BING2, DAP6, EAP1
UniGene
Hs.336916
Gene Chromosome Location
Chr.6: 33318558-33323016 on Build GRCh38
Transcripts
NM_001141969.1 NM_001141970.1 NM_001254717.1 NM_001350.4 XM_005248860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
85801 c.1976_1976delA p.N659fs*36 Chr.6: 33319184-33319184 on Build GRCh38
6969147 c.2045C>T p.S682F Chr.6: 33319115-33319115 on Build GRCh38
85802 c.1980_1980delG p.K662fs*33 Chr.6: 33319180-33319180 on Build GRCh38
4676233 c.2025C>A p.S675S Chr.6: 33319135-33319135 on Build GRCh38
3625706 c.2019G>A p.L673L Chr.6: 33319141-33319141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746856465 Chr.6: 33319195-33319195 on Build GRCh38
rs140525877 Chr.6: 33319129-33319129 on Build GRCh38
rs753556048 Chr.6: 33319130-33319130 on Build GRCh38
rs774763897 Chr.6: 33319227-33319227 on Build GRCh38
rs754567232 Chr.6: 33319137-33319137 on Build GRCh38
rs142786199 Chr.6: 33319146-33319146 on Build GRCh38
rs1801850 Chr.6: 33319178-33319178 on Build GRCh38
rs775384942 Chr.6: 33319207-33319207 on Build GRCh38
rs374737931 Chr.6: 33319172-33319172 on Build GRCh38
rs769949760 Chr.6: 33319196-33319196 on Build GRCh38
rs747956734 Chr.6: 33319170-33319170 on Build GRCh38
rs762148281 Chr.6: 33319234-33319234 on Build GRCh38
rs777866381 Chr.6: 33319186-33319186 on Build GRCh38
rs967799519 Chr.6: 33319182-33319182 on Build GRCh38
rs749571676 Chr.6: 33319214-33319214 on Build GRCh38
rs768834317 Chr.6: 33319216-33319216 on Build GRCh38
rs914779604 Chr.6: 33319165-33319165 on Build GRCh38

View Full Product Details