Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 45836508-45836656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID2
Entrez Gene ID
196528
Gene Name
AT-rich interaction domain 2
Gene Aliases
BAF200, p200
UniGene
Hs.317304
Gene Chromosome Location
Chr.12: 45729683-45908037 on Build GRCh38
Transcripts
NM_152641.2 XM_006719272.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
939495 c.739C>A p.R247S Chr.12: 45836622-45836622 on Build GRCh38
5414588 c.739C>T p.R247C Chr.12: 45836622-45836622 on Build GRCh38
5551912 c.728A>G p.D243G Chr.12: 45836611-45836611 on Build GRCh38
957512 c.772_772+20del21 p.? Chr.12: 45836655-45836655 on Build GRCh38
6951663 c.766T>G p.S256A Chr.12: 45836649-45836649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759197276 Chr.12: 45836548-45836548 on Build GRCh38
rs764704168 Chr.12: 45836550-45836550 on Build GRCh38
rs201425010 Chr.12: 45836553-45836553 on Build GRCh38
rs746113252 Chr.12: 45836576-45836576 on Build GRCh38
rs780015905 Chr.12: 45836585-45836585 on Build GRCh38
rs371094634 Chr.12: 45836556-45836556 on Build GRCh38
rs1019426458 Chr.12: 45836546-45836546 on Build GRCh38
rs186180564 Chr.12: 45836604-45836604 on Build GRCh38
rs756334486 Chr.12: 45836582-45836582 on Build GRCh38
rs766031938 Chr.12: 45836542-45836542 on Build GRCh38
rs199887651 Chr.12: 45836572-45836572 on Build GRCh38
rs751988354 Chr.12: 45836569-45836569 on Build GRCh38
rs753503220 Chr.12: 45836544-45836544 on Build GRCh38
rs975467320 Chr.12: 45836603-45836603 on Build GRCh38
rs757756425 Chr.12: 45836570-45836570 on Build GRCh38

View Full Product Details