Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 142458875-142459038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATR
Entrez Gene ID
545
Gene Name
ATR serine/threonine kinase
Gene Aliases
FCTCS, FRP1, MEC1, SCKL, SCKL1
UniGene
Hs.271791
Gene Chromosome Location
Chr.3: 142449235-142578826 on Build GRCh38
Transcripts
NM_001184.3 XM_011512924.1 XM_011512925.1 XM_017006643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6913613 c.7429C>T p.H2477Y Chr.3: 142459032-142459032 on Build GRCh38
7325212 c.7442T>A p.I2481N Chr.3: 142459019-142459019 on Build GRCh38
7246375 c.7503+10A>G p.? Chr.3: 142458948-142458948 on Build GRCh38
5350910 c.7459A>T p.T2487S Chr.3: 142459002-142459002 on Build GRCh38
6608479 c.7426C>T p.R2476C Chr.3: 142459035-142459035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373093280 Chr.3: 142458974-142458974 on Build GRCh38
rs767931735 Chr.3: 142458926-142458926 on Build GRCh38
rs369748890 Chr.3: 142458917-142458917 on Build GRCh38
rs574699024 Chr.3: 142458991-142458991 on Build GRCh38
rs556313656 Chr.3: 142458990-142458990 on Build GRCh38
rs368580370 Chr.3: 142458922-142458922 on Build GRCh38
rs758030592 Chr.3: 142458950-142458950 on Build GRCh38
rs764502964 Chr.3: 142458947-142458947 on Build GRCh38
rs752993414 Chr.3: 142458934-142458934 on Build GRCh38
rs750073869 Chr.3: 142458949-142458949 on Build GRCh38
rs376727616 Chr.3: 142458953-142458953 on Build GRCh38
rs751057775 Chr.3: 142458961-142458961 on Build GRCh38
rs761042012 Chr.3: 142458942-142458942 on Build GRCh38
rs369200535 Chr.3: 142458973-142458973 on Build GRCh38

View Full Product Details