Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 27952153-27952325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH2
Entrez Gene ID
1000
Gene Name
cadherin 2
Gene Aliases
CD325, CDHN, CDw325, NCAD
UniGene
Hs.464829
Gene Chromosome Location
Chr.18: 27932878-28177446 on Build GRCh38
Transcripts
NM_001308176.1 NM_001792.4 XM_011525788.1 XM_017025514.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1303646 c.2637T>G p.G879G Chr.18: 27952237-27952237 on Build GRCh38
385442 c.2678G>A p.R893Q Chr.18: 27952196-27952196 on Build GRCh38
3890839 c.2617C>T p.L873F Chr.18: 27952257-27952257 on Build GRCh38
6624776 c.2670G>T p.W890C Chr.18: 27952204-27952204 on Build GRCh38
386874 c.2673G>T p.G891G Chr.18: 27952201-27952201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776296541 Chr.18: 27952277-27952277 on Build GRCh38
rs765684003 Chr.18: 27952180-27952180 on Build GRCh38
rs375434529 Chr.18: 27952246-27952246 on Build GRCh38
rs770504867 Chr.18: 27952276-27952276 on Build GRCh38
rs201289431 Chr.18: 27952201-27952201 on Build GRCh38
rs879088485 Chr.18: 27952233-27952233 on Build GRCh38
rs756289185 Chr.18: 27952231-27952231 on Build GRCh38
rs200803866 Chr.18: 27952283-27952283 on Build GRCh38
rs759051449 Chr.18: 27952177-27952177 on Build GRCh38
rs200373316 Chr.18: 27952209-27952209 on Build GRCh38
rs201334347 Chr.18: 27952210-27952210 on Build GRCh38
rs764766275 Chr.18: 27952287-27952287 on Build GRCh38
rs753251005 Chr.18: 27952197-27952197 on Build GRCh38
rs377612781 Chr.18: 27952227-27952227 on Build GRCh38
rs896948558 Chr.18: 27952275-27952275 on Build GRCh38
rs771762304 Chr.18: 27952245-27952245 on Build GRCh38
rs764584049 Chr.18: 27952203-27952203 on Build GRCh38
rs201294768 Chr.18: 27952196-27952196 on Build GRCh38
rs201844490 Chr.18: 27952299-27952299 on Build GRCh38
rs773118735 Chr.18: 27952252-27952252 on Build GRCh38
rs776060409 Chr.18: 27952176-27952176 on Build GRCh38
rs138164198 Chr.18: 27952244-27952244 on Build GRCh38
rs746795245 Chr.18: 27952258-27952258 on Build GRCh38
rs199787442 Chr.18: 27952235-27952235 on Build GRCh38
rs199657884 Chr.18: 27952208-27952208 on Build GRCh38
rs758658207 Chr.18: 27952199-27952199 on Build GRCh38
rs371258872 Chr.18: 27952236-27952236 on Build GRCh38

View Full Product Details