Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 131981495-131981662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EP400
Entrez Gene ID
57634
Gene Name
E1A binding protein p400
Gene Aliases
CAGH32, P400, TNRC12
UniGene
Hs.723478
Gene Chromosome Location
Chr.12: 131949920-132080466 on Build GRCh38
Transcripts
NM_015409.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4681103 c.1503C>T p.D501D Chr.12: 131981556-131981556 on Build GRCh38
4528434 c.1645G>A p.A549T Chr.12: 131981590-131981590 on Build GRCh38
6462418 c.1509C>T p.G503G Chr.12: 131981562-131981562 on Build GRCh38
4843677 c.1454G>C p.R485P Chr.12: 131981507-131981507 on Build GRCh38
6406772 c.1628A>T p.Q543L Chr.12: 131981573-131981573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967160434 Chr.12: 131981605-131981605 on Build GRCh38
rs776276356 Chr.12: 131981619-131981619 on Build GRCh38
rs767712470 Chr.12: 131981567-131981567 on Build GRCh38
rs200663976 Chr.12: 131981563-131981563 on Build GRCh38
rs746379866 Chr.12: 131981625-131981625 on Build GRCh38
rs142075489 Chr.12: 131981522-131981522 on Build GRCh38
rs759672207 Chr.12: 131981530-131981530 on Build GRCh38
rs200859619 Chr.12: 131981585-131981585 on Build GRCh38
rs757930030 Chr.12: 131981597-131981597 on Build GRCh38
rs1034699546 Chr.12: 131981562-131981562 on Build GRCh38
rs775524582 Chr.12: 131981549-131981549 on Build GRCh38
rs555250384 Chr.12: 131981592-131981592 on Build GRCh38
rs936498578 Chr.12: 131981539-131981539 on Build GRCh38
rs569151190 Chr.12: 131981529-131981529 on Build GRCh38
rs563164907 Chr.12: 131981556-131981556 on Build GRCh38
rs754577236 Chr.12: 131981586-131981586 on Build GRCh38
rs375631970 Chr.12: 131981606-131981606 on Build GRCh38
rs906162275 Chr.12: 131981597-131981598 on Build GRCh38
rs149894039 Chr.12: 131981590-131981590 on Build GRCh38
rs769970714 Chr.12: 131981630-131981630 on Build GRCh38
rs770570595 Chr.12: 131981528-131981528 on Build GRCh38
rs1055278928 Chr.12: 131981626-131981626 on Build GRCh38
rs931620434 Chr.12: 131981635-131981635 on Build GRCh38
rs989992054 Chr.12: 131981589-131981589 on Build GRCh38
rs753596902 Chr.12: 131981577-131981577 on Build GRCh38
rs139101269 Chr.12: 131981553-131981553 on Build GRCh38
rs763493326 Chr.12: 131981552-131981552 on Build GRCh38
rs765414308 Chr.12: 131981544-131981544 on Build GRCh38
rs151135416 Chr.12: 131981527-131981527 on Build GRCh38
rs868109982 Chr.12: 131981584-131981584 on Build GRCh38
rs746850682 Chr.12: 131981611-131981611 on Build GRCh38
rs770826550 Chr.12: 131981613-131981613 on Build GRCh38
rs772401572 Chr.12: 131981637-131981637 on Build GRCh38
rs550529489 Chr.12: 131981621-131981622 on Build GRCh38

View Full Product Details