Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215361455-215361613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FN1
Entrez Gene ID
2335
Gene Name
fibronectin 1
Gene Aliases
CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
UniGene
Hs.203717
Gene Chromosome Location
Chr.2: 215360440-215436167 on Build GRCh38
Transcripts
NM_001306129.1 NM_001306130.1 NM_001306131.1 NM_001306132.1 NM_002026.3 NM_212474.2 NM_212476.2 NM_212478.2 NM_212482.2 XM_005246402.1 XM_005246405.1 XM_005246406.1 XM_005246409.1 XM_005246412.1 XM_005246414.1 XM_017003692.1 XM_017003693.1 XM_017003694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4091088 c.7060C>T p.R2354* Chr.2: 215361563-215361563 on Build GRCh38
3577498 c.7393C>T p.P2465S Chr.2: 215361596-215361596 on Build GRCh38
3673602 c.7060C>A p.R2354R Chr.2: 215361563-215361563 on Build GRCh38
4091087 c.7426C>T p.R2476* Chr.2: 215361563-215361563 on Build GRCh38
3426070 c.7040A>G p.Q2347R Chr.2: 215361583-215361583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756083637 Chr.2: 215361556-215361556 on Build GRCh38
rs966419080 Chr.2: 215361482-215361482 on Build GRCh38
rs146924311 Chr.2: 215361563-215361563 on Build GRCh38
rs771493768 Chr.2: 215361544-215361544 on Build GRCh38
rs778366340 Chr.2: 215361581-215361581 on Build GRCh38
rs1031803451 Chr.2: 215361496-215361496 on Build GRCh38
rs745904933 Chr.2: 215361526-215361526 on Build GRCh38
rs533318636 Chr.2: 215361486-215361486 on Build GRCh38
rs369527578 Chr.2: 215361562-215361562 on Build GRCh38
rs753002890 Chr.2: 215361550-215361550 on Build GRCh38
rs770103264 Chr.2: 215361541-215361541 on Build GRCh38
rs111587543 Chr.2: 215361523-215361523 on Build GRCh38
rs149294265 Chr.2: 215361587-215361587 on Build GRCh38
rs551557439 Chr.2: 215361549-215361549 on Build GRCh38
rs200851533 Chr.2: 215361552-215361552 on Build GRCh38
rs761834351 Chr.2: 215361528-215361529 on Build GRCh38
rs761308241 Chr.2: 215361530-215361530 on Build GRCh38
rs527756672 Chr.2: 215361576-215361576 on Build GRCh38
rs776879077 Chr.2: 215361517-215361518 on Build GRCh38
rs765471994 Chr.2: 215361547-215361548 on Build GRCh38
rs748495873 Chr.2: 215361515-215361515 on Build GRCh38
rs777514133 Chr.2: 215361549-215361551 on Build GRCh38
rs957522668 Chr.2: 215361534-215361534 on Build GRCh38
rs376904873 Chr.2: 215361545-215361545 on Build GRCh38
rs556448412 Chr.2: 215361550-215361551 on Build GRCh38
rs767405638 Chr.2: 215361551-215361551 on Build GRCh38
rs772528941 Chr.2: 215361521-215361521 on Build GRCh38

View Full Product Details