Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 52906091-52906259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCC
Entrez Gene ID
1630
Gene Name
DCC netrin 1 receptor
Gene Aliases
CRC18, CRCR1, IGDCC1, MRMV1, NTN1R1
UniGene
Hs.162025
Gene Chromosome Location
Chr.18: 52340172-53535903 on Build GRCh38
Transcripts
NM_005215.3 XM_011525843.1 XM_017025568.1 XM_017025569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6217084 c.45C>T p.P15P Chr.18: 52906132-52906132 on Build GRCh38
6488001 c.544C>A p.P182T Chr.18: 52906175-52906175 on Build GRCh38
5854250 c.166G>A p.G56R Chr.18: 52906253-52906253 on Build GRCh38
5205850 c.612delG p.D206fs*44 Chr.18: 52906243-52906243 on Build GRCh38
6524233 c.542C>A p.T181N Chr.18: 52906173-52906173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs797044552 Chr.18: 52906202-52906203 on Build GRCh38
rs868090609 Chr.18: 52906115-52906115 on Build GRCh38
rs200571296 Chr.18: 52906202-52906202 on Build GRCh38
rs200200224 Chr.18: 52906172-52906172 on Build GRCh38
rs559921087 Chr.18: 52906213-52906213 on Build GRCh38
rs756533345 Chr.18: 52906233-52906233 on Build GRCh38
rs121912967 Chr.18: 52906134-52906134 on Build GRCh38
rs1026050471 Chr.18: 52906171-52906171 on Build GRCh38
rs201360801 Chr.18: 52906201-52906201 on Build GRCh38
rs1042809360 Chr.18: 52906239-52906239 on Build GRCh38
rs372910352 Chr.18: 52906122-52906122 on Build GRCh38
rs750172123 Chr.18: 52906205-52906205 on Build GRCh38
rs915441598 Chr.18: 52906226-52906226 on Build GRCh38
rs746357250 Chr.18: 52906144-52906144 on Build GRCh38
rs964870675 Chr.18: 52906186-52906186 on Build GRCh38
rs776237905 Chr.18: 52906196-52906196 on Build GRCh38
rs768158522 Chr.18: 52906159-52906159 on Build GRCh38
rs138724679 Chr.18: 52906158-52906158 on Build GRCh38
rs760624202 Chr.18: 52906191-52906191 on Build GRCh38
rs199679918 Chr.18: 52906175-52906175 on Build GRCh38
rs771695804 Chr.18: 52906182-52906182 on Build GRCh38
rs767578049 Chr.18: 52906231-52906231 on Build GRCh38
rs757295777 Chr.18: 52906128-52906128 on Build GRCh38
rs201156075 Chr.18: 52906157-52906157 on Build GRCh38
rs751754292 Chr.18: 52906210-52906210 on Build GRCh38
rs377498043 Chr.18: 52906192-52906192 on Build GRCh38
rs572137094 Chr.18: 52906230-52906230 on Build GRCh38
rs2229080 Chr.18: 52906232-52906232 on Build GRCh38
rs772925573 Chr.18: 52906184-52906184 on Build GRCh38
rs781294118 Chr.18: 52906142-52906142 on Build GRCh38

View Full Product Details