Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 3153889-3154057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRBN
Entrez Gene ID
51185
Gene Name
cereblon
Gene Aliases
MRT2, MRT2A
UniGene
Hs.18925
Gene Chromosome Location
Chr.3: 3148490-3179717 on Build GRCh38
Transcripts
NM_001173482.1 NM_016302.3 XM_005265202.3 XM_011533791.2 XM_011533793.2 XM_011533794.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792946 c.951+10G>A p.? Chr.3: 3153950-3153950 on Build GRCh38
1692303 c.895C>T p.L299F Chr.3: 3154016-3154016 on Build GRCh38
4382150 c.920G>A p.R307Q Chr.3: 3153991-3153991 on Build GRCh38
3427514 c.884G>T p.R295I Chr.3: 3154027-3154027 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758001907 Chr.3: 3154009-3154009 on Build GRCh38
rs1053571121 Chr.3: 3153981-3153981 on Build GRCh38
rs529742781 Chr.3: 3153931-3153931 on Build GRCh38
rs774753303 Chr.3: 3153943-3153944 on Build GRCh38
rs745521075 Chr.3: 3153988-3153988 on Build GRCh38
rs755466110 Chr.3: 3153934-3153934 on Build GRCh38
rs142337178 Chr.3: 3153974-3153974 on Build GRCh38
rs181497249 Chr.3: 3153924-3153924 on Build GRCh38
rs756804291 Chr.3: 3153965-3153965 on Build GRCh38
rs779833529 Chr.3: 3154029-3154029 on Build GRCh38
rs751880211 Chr.3: 3153929-3153929 on Build GRCh38
rs375361013 Chr.3: 3154001-3154001 on Build GRCh38
rs768167718 Chr.3: 3153946-3153946 on Build GRCh38
rs549822663 Chr.3: 3153957-3153957 on Build GRCh38
rs752048403 Chr.3: 3153991-3153991 on Build GRCh38

View Full Product Details