Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180603055-180603222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT4
Entrez Gene ID
2324
Gene Name
fms related tyrosine kinase 4
Gene Aliases
FLT-4, FLT41, LMPH1A, PCL, VEGFR-3, VEGFR3
UniGene
Hs.646917
Gene Chromosome Location
Chr.5: 180601506-180650298 on Build GRCh38
Transcripts
NM_182925.4 XM_011534478.2 XM_011534484.2 XM_017009263.1 XM_017009268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6665374 c.4085G>T p.S1362I Chr.5: 180603199-180603199 on Build GRCh38
4685052 c.4063G>A p.V1355M Chr.5: 180603221-180603221 on Build GRCh38
3947347 c.4062C>A p.R1354R Chr.5: 180603222-180603222 on Build GRCh38
5403228 c.4062C>G p.R1354R Chr.5: 180603222-180603222 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs900565725 Chr.5: 180603124-180603124 on Build GRCh38
rs776319807 Chr.5: 180603146-180603146 on Build GRCh38
rs753977164 Chr.5: 180603165-180603165 on Build GRCh38
rs768618179 Chr.5: 180603145-180603145 on Build GRCh38
rs199750262 Chr.5: 180603183-180603183 on Build GRCh38
rs768540740 Chr.5: 180603197-180603197 on Build GRCh38
rs761402024 Chr.5: 180603149-180603149 on Build GRCh38
rs201631586 Chr.5: 180603193-180603193 on Build GRCh38
rs1004628438 Chr.5: 180603098-180603098 on Build GRCh38
rs765039293 Chr.5: 180603150-180603150 on Build GRCh38
rs373394049 Chr.5: 180603141-180603141 on Build GRCh38
rs757342606 Chr.5: 180603169-180603169 on Build GRCh38
rs767800556 Chr.5: 180603157-180603157 on Build GRCh38
rs377342113 Chr.5: 180603142-180603142 on Build GRCh38
rs746111269 Chr.5: 180603177-180603177 on Build GRCh38
rs762595641 Chr.5: 180603151-180603151 on Build GRCh38
rs113034305 Chr.5: 180603132-180603132 on Build GRCh38
rs753054153 Chr.5: 180603159-180603159 on Build GRCh38
rs199812896 Chr.5: 180603175-180603175 on Build GRCh38
rs756621538 Chr.5: 180603162-180603162 on Build GRCh38
rs190791214 Chr.5: 180603184-180603184 on Build GRCh38
rs370516958 Chr.5: 180603163-180603163 on Build GRCh38

View Full Product Details