Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 45731268-45731415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID2
Entrez Gene ID
196528
Gene Name
AT-rich interaction domain 2
Gene Aliases
BAF200, p200
UniGene
Hs.317304
Gene Chromosome Location
Chr.12: 45729683-45908037 on Build GRCh38
Transcripts
NM_152641.2 XM_006719272.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7403527 c.281_282insA p.R95fs*16 Chr.12: 45731311-45731312 on Build GRCh38
3460959 c.284+1G>A p.? Chr.12: 45731315-45731315 on Build GRCh38
5951321 c.284+4_284+5insT p.? Chr.12: 45731318-45731319 on Build GRCh38
6940580 c.281_282insG p.R95fs*16 Chr.12: 45731311-45731312 on Build GRCh38
5361621 c.246T>C p.C82C Chr.12: 45731276-45731276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915751950 Chr.12: 45731357-45731357 on Build GRCh38
rs760167654 Chr.12: 45731323-45731323 on Build GRCh38
rs777255828 Chr.12: 45731336-45731336 on Build GRCh38
rs1039864019 Chr.12: 45731310-45731310 on Build GRCh38
rs192612373 Chr.12: 45731385-45731385 on Build GRCh38
rs764259718 Chr.12: 45731350-45731350 on Build GRCh38
rs763186568 Chr.12: 45731349-45731349 on Build GRCh38
rs948559499 Chr.12: 45731360-45731360 on Build GRCh38
rs772882172 Chr.12: 45731305-45731305 on Build GRCh38
rs771359605 Chr.12: 45731335-45731335 on Build GRCh38
rs902161806 Chr.12: 45731390-45731390 on Build GRCh38
rs753876665 Chr.12: 45731365-45731365 on Build GRCh38
rs368682134 Chr.12: 45731382-45731382 on Build GRCh38
rs765630775 Chr.12: 45731339-45731339 on Build GRCh38
rs751593644 Chr.12: 45731361-45731361 on Build GRCh38
rs371802419 Chr.12: 45731346-45731346 on Build GRCh38

View Full Product Details