Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215370188-215370335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FN1
Entrez Gene ID
2335
Gene Name
fibronectin 1
Gene Aliases
CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
UniGene
Hs.203717
Gene Chromosome Location
Chr.2: 215360440-215436167 on Build GRCh38
Transcripts
NM_001306129.1 NM_001306130.1 NM_001306131.1 NM_001306132.1 NM_002026.3 NM_212474.2 NM_212476.2 NM_212478.2 NM_212482.2 XM_005246397.1 XM_005246398.1 XM_005246399.1 XM_005246401.1 XM_005246402.1 XM_005246403.1 XM_005246404.1 XM_005246405.1 XM_005246406.1 XM_005246407.1 XM_005246408.1 XM_005246409.1 XM_005246410.1 XM_005246411.1 XM_005246412.1 XM_005246414.1 XM_005246416.1 XM_017003692.1 XM_017003693.1 XM_017003694.1 XM_017003695.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4685180 c.6475G>A p.V2159M Chr.2: 215370306-215370306 on Build GRCh38
4901003 c.6451A>G p.K2151E Chr.2: 215370330-215370330 on Build GRCh38
7368573 c.6851C>G p.S2284C Chr.2: 215370296-215370296 on Build GRCh38
4685179 c.6841G>A p.V2281M Chr.2: 215370306-215370306 on Build GRCh38
4901002 c.6817A>G p.K2273E Chr.2: 215370330-215370330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111303746 Chr.2: 215370282-215370282 on Build GRCh38
rs758735728 Chr.2: 215370276-215370276 on Build GRCh38
rs984314133 Chr.2: 215370215-215370215 on Build GRCh38
rs756035028 Chr.2: 215370246-215370246 on Build GRCh38
rs114199884 Chr.2: 215370265-215370265 on Build GRCh38
rs199573501 Chr.2: 215370264-215370264 on Build GRCh38
rs55706546 Chr.2: 215370306-215370306 on Build GRCh38
rs1039670299 Chr.2: 215370241-215370241 on Build GRCh38
rs759556957 Chr.2: 215370273-215370273 on Build GRCh38
rs55739979 Chr.2: 215370258-215370258 on Build GRCh38
rs753281522 Chr.2: 215370263-215370263 on Build GRCh38
rs375230206 Chr.2: 215370266-215370266 on Build GRCh38
rs910113386 Chr.2: 215370218-215370218 on Build GRCh38
rs770084828 Chr.2: 215370307-215370307 on Build GRCh38
rs753140099 Chr.2: 215370267-215370267 on Build GRCh38
rs990526466 Chr.2: 215370308-215370308 on Build GRCh38
rs757901031 Chr.2: 215370311-215370311 on Build GRCh38
rs1000352801 Chr.2: 215370287-215370287 on Build GRCh38
rs769831640 Chr.2: 215370269-215370270 on Build GRCh38
rs527529776 Chr.2: 215370243-215370243 on Build GRCh38
rs200636538 Chr.2: 215370281-215370281 on Build GRCh38
rs1043555323 Chr.2: 215370295-215370295 on Build GRCh38
rs111364075 Chr.2: 215370293-215370293 on Build GRCh38
rs781433774 Chr.2: 215370297-215370297 on Build GRCh38

View Full Product Details