Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 112231559-112231691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD3
Entrez Gene ID
114788
Gene Name
CUB and Sushi multiple domains 3
Gene Aliases
-
UniGene
Hs.91381
Gene Chromosome Location
Chr.8: 112222928-113437499 on Build GRCh38
Transcripts
NM_052900.2 NM_198123.1 NM_198124.1 XM_011516815.2 XM_011516816.2 XM_017013008.1 XM_017013009.1 XM_017013010.1 XM_017013011.1 XM_017013012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1137795 c.10661G>T p.G3554V Chr.8: 112231592-112231592 on Build GRCh38
4979534 c.10683T>C p.Y3561Y Chr.8: 112231570-112231570 on Build GRCh38
378543 c.10741-1G>T p.? Chr.8: 112231633-112231633 on Build GRCh38
201485 c.10798G>T p.D3600Y Chr.8: 112231575-112231575 on Build GRCh38
1249302 c.10797A>C p.K3599N Chr.8: 112231576-112231576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376588062 Chr.8: 112231667-112231667 on Build GRCh38
rs751012975 Chr.8: 112231653-112231653 on Build GRCh38
rs766890042 Chr.8: 112231636-112231636 on Build GRCh38
rs1056178539 Chr.8: 112231621-112231621 on Build GRCh38
rs772298520 Chr.8: 112231602-112231602 on Build GRCh38
rs748183238 Chr.8: 112231588-112231588 on Build GRCh38
rs777273246 Chr.8: 112231645-112231645 on Build GRCh38
rs773590013 Chr.8: 112231603-112231603 on Build GRCh38
rs756745702 Chr.8: 112231663-112231663 on Build GRCh38
rs761096772 Chr.8: 112231635-112231635 on Build GRCh38
rs1001474038 Chr.8: 112231613-112231613 on Build GRCh38
rs904562910 Chr.8: 112231587-112231587 on Build GRCh38

View Full Product Details