Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 75029318-75029479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUPD1
Entrez Gene ID
338599
Gene Name
dual specificity phosphatase and pro isomerase domain containing 1
Gene Aliases
DUSP27, FMDSP
UniGene
Hs.535090
Gene Chromosome Location
Chr.10: 75026427-75073827 on Build GRCh38
Transcripts
XM_011539747.2 NM_001256468.1 NM_001256469.1 NM_012330.3 XM_005269664.2 XM_017016000.1 XM_017016001.1 XM_017016002.1 XM_017016003.1 XM_017016004.1 XM_017016005.1 XM_017016006.1 XM_017016007.1 XM_017016008.1 XM_017016009.1

Gene Symbol
KAT6B
Entrez Gene ID
23522
Gene Name
lysine acetyltransferase 6B
Gene Aliases
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf
UniGene
Hs.35758
Gene Chromosome Location
Chr.10: 74824927-75032623 on Build GRCh38
Transcripts
XM_011539747.2 NM_001256468.1 NM_001256469.1 NM_012330.3 XM_005269664.2 XM_017016000.1 XM_017016001.1 XM_017016002.1 XM_017016003.1 XM_017016004.1 XM_017016005.1 XM_017016006.1 XM_017016007.1 XM_017016008.1 XM_017016009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5576215 c.4622C>G p.T1541S Chr.10: 75029446-75029446 on Build GRCh38
1965163 c.4569C>T p.V1523V Chr.10: 75029393-75029393 on Build GRCh38
3730496 c.4632C>T p.A1544A Chr.10: 75029456-75029456 on Build GRCh38
240809 c.4501G>T p.E1501* Chr.10: 75029325-75029325 on Build GRCh38
4694066 c.4611C>T p.I1537I Chr.10: 75029435-75029435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760322602 Chr.10: 75029391-75029391 on Build GRCh38
rs753937127 Chr.10: 75029436-75029436 on Build GRCh38
rs765058663 Chr.10: 75029411-75029411 on Build GRCh38
rs779545571 Chr.10: 75029422-75029424 on Build GRCh38
rs772508131 Chr.10: 75029358-75029358 on Build GRCh38
rs750426572 Chr.10: 75029414-75029414 on Build GRCh38
rs758083734 Chr.10: 75029420-75029420 on Build GRCh38
rs545973592 Chr.10: 75029394-75029394 on Build GRCh38
rs752901942 Chr.10: 75029427-75029427 on Build GRCh38
rs766230039 Chr.10: 75029426-75029426 on Build GRCh38
rs772255130 Chr.10: 75029456-75029456 on Build GRCh38
rs375290304 Chr.10: 75029453-75029453 on Build GRCh38
rs1014954135 Chr.10: 75029347-75029347 on Build GRCh38
rs746194410 Chr.10: 75029354-75029354 on Build GRCh38
rs371945178 Chr.10: 75029435-75029435 on Build GRCh38
rs572347318 Chr.10: 75029360-75029360 on Build GRCh38
rs761383444 Chr.10: 75029406-75029406 on Build GRCh38
rs771559131 Chr.10: 75029346-75029346 on Build GRCh38
rs920418680 Chr.10: 75029397-75029397 on Build GRCh38
rs774963101 Chr.10: 75029365-75029365 on Build GRCh38
rs779236437 Chr.10: 75029447-75029447 on Build GRCh38
rs746334369 Chr.10: 75029348-75029350 on Build GRCh38
rs774962648 Chr.10: 75029353-75029353 on Build GRCh38
rs145158232 Chr.10: 75029457-75029457 on Build GRCh38
rs367789559 Chr.10: 75029399-75029399 on Build GRCh38
rs757461393 Chr.10: 75029441-75029441 on Build GRCh38

View Full Product Details