Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 77520772-77520918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRX
Entrez Gene ID
546
Gene Name
ATRX, chromatin remodeler
Gene Aliases
ATR2, JMS, MRX52, MRXHF1, RAD54, RAD54L, SFM1, SHS, XH2, XNP, ZNF-HX
UniGene
Hs.533526
Gene Chromosome Location
Chr.X: 77504878-77786269 on Build GRCh38
Transcripts
NM_000489.4 NM_138270.3 XM_005262153.4 XM_005262154.4 XM_005262155.4 XM_005262156.4 XM_005262157.4 XM_006724666.3 XM_006724667.3 XM_017029601.1 XM_017029602.1 XM_017029603.1 XM_017029604.1 XM_017029605.1 XM_017029606.1 XM_017029607.1 XM_017029608.1 XM_017029609.1 XM_017029610.1 XM_017029611.1

SNP Detail

SNP ID Chromosome Location
rs782703242 Chr.X: 77520875-77520875 on Build GRCh38
rs1057518225 Chr.X: 77520841-77520841 on Build GRCh38
rs112385254 Chr.X: 77520815-77520815 on Build GRCh38
rs1049073136 Chr.X: 77520820-77520820 on Build GRCh38
rs122445100 Chr.X: 77520826-77520826 on Build GRCh38
rs122445099 Chr.X: 77520832-77520832 on Build GRCh38
rs782490070 Chr.X: 77520805-77520805 on Build GRCh38
rs938190596 Chr.X: 77520806-77520806 on Build GRCh38
rs147341551 Chr.X: 77520877-77520877 on Build GRCh38

View Full Product Details