Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215361563-215361732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FN1
Entrez Gene ID
2335
Gene Name
fibronectin 1
Gene Aliases
CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
UniGene
Hs.203717
Gene Chromosome Location
Chr.2: 215360440-215436167 on Build GRCh38
Transcripts
NM_001306129.1 NM_001306130.1 NM_001306131.1 NM_001306132.1 NM_002026.3 NM_212474.2 NM_212476.2 NM_212478.2 NM_212482.2 XM_005246402.1 XM_005246405.1 XM_005246406.1 XM_005246409.1 XM_005246412.1 XM_005246414.1 XM_017003692.1 XM_017003693.1 XM_017003694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1153730 c.7373G>A p.C2458Y Chr.2: 215361616-215361616 on Build GRCh38
4091088 c.7060C>T p.R2354* Chr.2: 215361563-215361563 on Build GRCh38
3577498 c.7393C>T p.P2465S Chr.2: 215361596-215361596 on Build GRCh38
3673602 c.7060C>A p.R2354R Chr.2: 215361563-215361563 on Build GRCh38
1016153 c.7007G>A p.C2336Y Chr.2: 215361616-215361616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749150808 Chr.2: 215361691-215361691 on Build GRCh38
rs752005289 Chr.2: 215361645-215361646 on Build GRCh38
rs183520841 Chr.2: 215361672-215361672 on Build GRCh38
rs574336108 Chr.2: 215361621-215361621 on Build GRCh38
rs770041184 Chr.2: 215361620-215361620 on Build GRCh38
rs763371667 Chr.2: 215361631-215361632 on Build GRCh38
rs188055354 Chr.2: 215361695-215361695 on Build GRCh38
rs777236760 Chr.2: 215361598-215361598 on Build GRCh38
rs201347712 Chr.2: 215361638-215361638 on Build GRCh38
rs149294265 Chr.2: 215361587-215361587 on Build GRCh38
rs747275525 Chr.2: 215361591-215361591 on Build GRCh38
rs889680092 Chr.2: 215361696-215361696 on Build GRCh38
rs763302431 Chr.2: 215361632-215361632 on Build GRCh38
rs116671379 Chr.2: 215361656-215361656 on Build GRCh38
rs746344265 Chr.2: 215361615-215361615 on Build GRCh38
rs944525749 Chr.2: 215361606-215361606 on Build GRCh38
rs35258787 Chr.2: 215361694-215361695 on Build GRCh38
rs539682987 Chr.2: 215361657-215361657 on Build GRCh38
rs760416846 Chr.2: 215361642-215361642 on Build GRCh38
rs752078579 Chr.2: 215361655-215361655 on Build GRCh38
rs771587539 Chr.2: 215361597-215361597 on Build GRCh38
rs757979640 Chr.2: 215361588-215361588 on Build GRCh38
rs112429571 Chr.2: 215361682-215361683 on Build GRCh38
rs777238193 Chr.2: 215361669-215361669 on Build GRCh38
rs781672930 Chr.2: 215361677-215361677 on Build GRCh38
rs76784243 Chr.2: 215361651-215361651 on Build GRCh38
rs751678344 Chr.2: 215361694-215361694 on Build GRCh38
rs778200057 Chr.2: 215361589-215361589 on Build GRCh38
rs773073457 Chr.2: 215361641-215361641 on Build GRCh38
rs201573768 Chr.2: 215361645-215361645 on Build GRCh38
rs550551506 Chr.2: 215361652-215361652 on Build GRCh38
rs113068773 Chr.2: 215361670-215361670 on Build GRCh38
rs961174901 Chr.2: 215361682-215361682 on Build GRCh38
rs766732265 Chr.2: 215361646-215361646 on Build GRCh38
rs764866446 Chr.2: 215361654-215361654 on Build GRCh38
rs755503314 Chr.2: 215361653-215361655 on Build GRCh38

View Full Product Details