Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 75028557-75028728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUPD1
Entrez Gene ID
338599
Gene Name
dual specificity phosphatase and pro isomerase domain containing 1
Gene Aliases
DUSP27, FMDSP
UniGene
Hs.535090
Gene Chromosome Location
Chr.10: 75026427-75073827 on Build GRCh38
Transcripts
XM_011539747.2 NM_001256468.1 NM_001256469.1 NM_012330.3 XM_005269664.2 XM_017016000.1 XM_017016001.1 XM_017016002.1 XM_017016003.1 XM_017016004.1 XM_017016005.1 XM_017016006.1 XM_017016007.1 XM_017016008.1 XM_017016009.1

Gene Symbol
KAT6B
Entrez Gene ID
23522
Gene Name
lysine acetyltransferase 6B
Gene Aliases
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf
UniGene
Hs.35758
Gene Chromosome Location
Chr.10: 74824927-75032623 on Build GRCh38
Transcripts
XM_011539747.2 NM_001256468.1 NM_001256469.1 NM_012330.3 XM_005269664.2 XM_017016000.1 XM_017016001.1 XM_017016002.1 XM_017016003.1 XM_017016004.1 XM_017016005.1 XM_017016006.1 XM_017016007.1 XM_017016008.1 XM_017016009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
212182 c.3761G>A p.R1254H Chr.10: 75028585-75028585 on Build GRCh38
4930687 c.3831A>T p.P1277P Chr.10: 75028655-75028655 on Build GRCh38
7248328 c.3744G>T p.W1248C Chr.10: 75028568-75028568 on Build GRCh38
7145527 c.3882G>C p.E1294D Chr.10: 75028706-75028706 on Build GRCh38
5942332 c.3752G>A p.G1251E Chr.10: 75028576-75028576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375206291 Chr.10: 75028625-75028625 on Build GRCh38
rs141935235 Chr.10: 75028652-75028652 on Build GRCh38
rs371467644 Chr.10: 75028592-75028592 on Build GRCh38
rs759027870 Chr.10: 75028619-75028619 on Build GRCh38
rs756421689 Chr.10: 75028679-75028679 on Build GRCh38
rs766315089 Chr.10: 75028661-75028661 on Build GRCh38
rs147574112 Chr.10: 75028587-75028587 on Build GRCh38
rs376367047 Chr.10: 75028676-75028676 on Build GRCh38
rs368121049 Chr.10: 75028663-75028663 on Build GRCh38
rs577563268 Chr.10: 75028664-75028664 on Build GRCh38
rs371747862 Chr.10: 75028671-75028671 on Build GRCh38
rs978272228 Chr.10: 75028653-75028653 on Build GRCh38
rs568525923 Chr.10: 75028585-75028585 on Build GRCh38
rs539703139 Chr.10: 75028648-75028648 on Build GRCh38
rs752632829 Chr.10: 75028645-75028645 on Build GRCh38
rs199470470 Chr.10: 75028593-75028596 on Build GRCh38
rs770280505 Chr.10: 75028595-75028595 on Build GRCh38
rs772133401 Chr.10: 75028620-75028620 on Build GRCh38
rs1028624445 Chr.10: 75028614-75028614 on Build GRCh38
rs763998844 Chr.10: 75028629-75028629 on Build GRCh38
rs770422861 Chr.10: 75028593-75028595 on Build GRCh38
rs770307489 Chr.10: 75028665-75028665 on Build GRCh38
rs199470473 Chr.10: 75028626-75028626 on Build GRCh38
rs199470472 Chr.10: 75028612-75028613 on Build GRCh38
rs777272076 Chr.10: 75028658-75028658 on Build GRCh38
rs199470471 Chr.10: 75028597-75028598 on Build GRCh38
rs145119505 Chr.10: 75028586-75028586 on Build GRCh38
rs572647460 Chr.10: 75028670-75028670 on Build GRCh38
rs199470474 Chr.10: 75028701-75028701 on Build GRCh38
rs988830424 Chr.10: 75028583-75028583 on Build GRCh38
rs755119518 Chr.10: 75028666-75028666 on Build GRCh38
rs565994476 Chr.10: 75028624-75028624 on Build GRCh38
rs773926507 Chr.10: 75028604-75028604 on Build GRCh38

View Full Product Details