Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 142449445-142449612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATR
Entrez Gene ID
545
Gene Name
ATR serine/threonine kinase
Gene Aliases
FCTCS, FRP1, MEC1, SCKL, SCKL1
UniGene
Hs.271791
Gene Chromosome Location
Chr.3: 142449235-142578826 on Build GRCh38
Transcripts
NM_001184.3 XM_011512924.1 XM_011512925.1 XM_017006643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5625667 c.7841T>C p.L2614S Chr.3: 142449523-142449523 on Build GRCh38
5006683 c.7842A>C p.L2614F Chr.3: 142449522-142449522 on Build GRCh38
4436622 c.7798C>A p.Q2600K Chr.3: 142449566-142449566 on Build GRCh38
6935124 c.7916G>A p.G2639D Chr.3: 142449448-142449448 on Build GRCh38
1039339 c.7777C>A p.L2593I Chr.3: 142449587-142449587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368061331 Chr.3: 142449507-142449507 on Build GRCh38
rs374127772 Chr.3: 142449500-142449500 on Build GRCh38
rs1047970453 Chr.3: 142449567-142449567 on Build GRCh38
rs781420658 Chr.3: 142449506-142449506 on Build GRCh38
rs749753899 Chr.3: 142449548-142449548 on Build GRCh38
rs371176601 Chr.3: 142449525-142449525 on Build GRCh38
rs1034620994 Chr.3: 142449526-142449526 on Build GRCh38
rs199948706 Chr.3: 142449547-142449547 on Build GRCh38
rs958635262 Chr.3: 142449534-142449534 on Build GRCh38
rs767609266 Chr.3: 142449539-142449540 on Build GRCh38
rs1802904 Chr.3: 142449489-142449489 on Build GRCh38
rs768358025 Chr.3: 142449578-142449578 on Build GRCh38
rs112027460 Chr.3: 142449485-142449485 on Build GRCh38
rs771519566 Chr.3: 142449553-142449553 on Build GRCh38
rs750316781 Chr.3: 142449474-142449474 on Build GRCh38
rs774810249 Chr.3: 142449555-142449555 on Build GRCh38
rs758432141 Chr.3: 142449479-142449479 on Build GRCh38
rs765167556 Chr.3: 142449473-142449473 on Build GRCh38
rs746266532 Chr.3: 142449564-142449564 on Build GRCh38

View Full Product Details