Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 142864369-142864542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB6
Entrez Gene ID
2051
Gene Name
EPH receptor B6
Gene Aliases
HEP
UniGene
Hs.380089
Gene Chromosome Location
Chr.7: 142854640-142871093 on Build GRCh38
Transcripts
NM_001280794.2 NM_001280795.2 NM_004445.5 NR_104001.2 XM_006715881.2 XM_011515879.1 XM_011515880.1 XM_011515881.2 XM_011515882.2

SNP Detail

SNP ID Chromosome Location
rs148221019 Chr.7: 142864505-142864505 on Build GRCh38
rs541042559 Chr.7: 142864472-142864472 on Build GRCh38
rs375814622 Chr.7: 142864388-142864388 on Build GRCh38
rs780569137 Chr.7: 142864492-142864492 on Build GRCh38
rs774677941 Chr.7: 142864439-142864439 on Build GRCh38
rs755527525 Chr.7: 142864487-142864487 on Build GRCh38
rs762082450 Chr.7: 142864449-142864449 on Build GRCh38
rs55742403 Chr.7: 142864465-142864465 on Build GRCh38
rs780639428 Chr.7: 142864481-142864481 on Build GRCh38
rs766413236 Chr.7: 142864512-142864512 on Build GRCh38
rs746083143 Chr.7: 142864450-142864451 on Build GRCh38
rs149090003 Chr.7: 142864399-142864399 on Build GRCh38
rs767628990 Chr.7: 142864450-142864450 on Build GRCh38
rs751382944 Chr.7: 142864473-142864473 on Build GRCh38
rs754136264 Chr.7: 142864467-142864467 on Build GRCh38
rs143064665 Chr.7: 142864424-142864424 on Build GRCh38
rs751082954 Chr.7: 142864451-142864451 on Build GRCh38
rs377242493 Chr.7: 142864506-142864506 on Build GRCh38
rs369872682 Chr.7: 142864398-142864398 on Build GRCh38
rs373135466 Chr.7: 142864484-142864484 on Build GRCh38
rs866141009 Chr.7: 142864446-142864446 on Build GRCh38
rs1005868917 Chr.7: 142864423-142864423 on Build GRCh38
rs756998208 Chr.7: 142864476-142864476 on Build GRCh38
rs765246993 Chr.7: 142864454-142864454 on Build GRCh38
rs768967254 Chr.7: 142864425-142864425 on Build GRCh38
rs555319297 Chr.7: 142864417-142864417 on Build GRCh38
rs769138939 Chr.7: 142864494-142864494 on Build GRCh38
rs752646274 Chr.7: 142864455-142864455 on Build GRCh38
rs774764025 Chr.7: 142864497-142864497 on Build GRCh38
rs770338059 Chr.7: 142864403-142864403 on Build GRCh38
rs746695848 Chr.7: 142864393-142864393 on Build GRCh38
rs200894619 Chr.7: 142864389-142864389 on Build GRCh38
rs756835314 Chr.7: 142864396-142864396 on Build GRCh38
rs760618193 Chr.7: 142864452-142864452 on Build GRCh38

View Full Product Details