Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7393558-7393789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLSCR3
Entrez Gene ID
57048
Gene Name
phospholipid scramblase 3
Gene Aliases
-
UniGene
Hs.534591
Gene Chromosome Location
Chr.17: 7389727-7394843 on Build GRCh38
Transcripts
NM_001201576.1 NM_020360.3 NR_037719.1

Gene Symbol
TMEM256-PLSCR3
Entrez Gene ID
100529211
Gene Name
TMEM256-PLSCR3 readthrough (NMD candidate)
Gene Aliases
C17orf61-PLSCR3
UniGene
Hs.534591
Gene Chromosome Location
Chr.17: 7389727-7404131 on Build GRCh38
Transcripts
NM_001201576.1 NM_020360.3 NR_037719.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5972403 c.360C>T p.F120F Chr.17: 7393697-7393697 on Build GRCh38
263006 c.229G>A p.E77K Chr.17: 7393615-7393615 on Build GRCh38
3356959 c.145T>C p.F49L Chr.17: 7393699-7393699 on Build GRCh38
5468201 c.151C>T p.L51F Chr.17: 7393693-7393693 on Build GRCh38
6403936 c.360C>G p.F120L Chr.17: 7393697-7393697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778973104 Chr.17: 7393622-7393622 on Build GRCh38
rs748255530 Chr.17: 7393650-7393650 on Build GRCh38
rs1024280923 Chr.17: 7393668-7393668 on Build GRCh38
rs72842811 Chr.17: 7393580-7393580 on Build GRCh38
rs748174964 Chr.17: 7393765-7393765 on Build GRCh38
rs200961730 Chr.17: 7393607-7393607 on Build GRCh38
rs561101958 Chr.17: 7393628-7393628 on Build GRCh38
rs760298352 Chr.17: 7393688-7393688 on Build GRCh38
rs757131492 Chr.17: 7393620-7393620 on Build GRCh38
rs373262490 Chr.17: 7393744-7393744 on Build GRCh38
rs963930094 Chr.17: 7393581-7393581 on Build GRCh38
rs773554189 Chr.17: 7393661-7393661 on Build GRCh38
rs752136675 Chr.17: 7393687-7393687 on Build GRCh38
rs769844028 Chr.17: 7393651-7393651 on Build GRCh38
rs772167905 Chr.17: 7393636-7393636 on Build GRCh38
rs201923475 Chr.17: 7393746-7393746 on Build GRCh38
rs753578165 Chr.17: 7393708-7393708 on Build GRCh38
rs756972692 Chr.17: 7393724-7393724 on Build GRCh38
rs768024448 Chr.17: 7393679-7393679 on Build GRCh38
rs199962387 Chr.17: 7393615-7393615 on Build GRCh38
rs759983478 Chr.17: 7393677-7393677 on Build GRCh38
rs755663748 Chr.17: 7393593-7393593 on Build GRCh38
rs908645011 Chr.17: 7393736-7393736 on Build GRCh38
rs761754400 Chr.17: 7393633-7393633 on Build GRCh38
rs992982425 Chr.17: 7393743-7393743 on Build GRCh38
rs778811511 Chr.17: 7393726-7393726 on Build GRCh38
rs371969579 Chr.17: 7393739-7393739 on Build GRCh38
rs766969335 Chr.17: 7393584-7393584 on Build GRCh38
rs973943878 Chr.17: 7393747-7393747 on Build GRCh38
rs780148424 Chr.17: 7393764-7393764 on Build GRCh38
rs765324845 Chr.17: 7393673-7393673 on Build GRCh38
rs750266647 Chr.17: 7393745-7393745 on Build GRCh38
rs766804593 Chr.17: 7393672-7393672 on Build GRCh38
rs908648200 Chr.17: 7393693-7393693 on Build GRCh38
rs368081936 Chr.17: 7393592-7393592 on Build GRCh38
rs948934055 Chr.17: 7393768-7393768 on Build GRCh38
rs758357139 Chr.17: 7393761-7393761 on Build GRCh38
rs531762972 Chr.17: 7393649-7393649 on Build GRCh38
rs549970908 Chr.17: 7393705-7393705 on Build GRCh38
rs763202142 Chr.17: 7393667-7393667 on Build GRCh38

View Full Product Details