Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 105567132-105567365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB10
Entrez Gene ID
100130771
Gene Name
EF-hand calcium binding domain 10
Gene Aliases
-
UniGene
Hs.719432
Gene Chromosome Location
Chr.7: 105565133-105581529 on Build GRCh38
Transcripts
NR_027068.1 NM_021930.4 XM_005250524.3 XM_011516458.2 XM_017012496.1 XM_017012497.1 XM_017012498.1

Gene Symbol
RINT1
Entrez Gene ID
60561
Gene Name
RAD50 interactor 1
Gene Aliases
RINT-1
UniGene
Hs.531388
Gene Chromosome Location
Chr.7: 105532085-105567688 on Build GRCh38
Transcripts
NR_027068.1 NM_021930.4 XM_005250524.3 XM_011516458.2 XM_017012496.1 XM_017012497.1 XM_017012498.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4384181 c.2334G>C p.E778D Chr.7: 105567266-105567266 on Build GRCh38
6766809 c.2224G>A p.G742S Chr.7: 105567156-105567156 on Build GRCh38
4384180 c.2288C>T p.A763V Chr.7: 105567220-105567220 on Build GRCh38
4759747 c.2310C>A p.Y770* Chr.7: 105567242-105567242 on Build GRCh38
4161702 c.2314C>A p.L772M Chr.7: 105567246-105567246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760695096 Chr.7: 105567185-105567185 on Build GRCh38
rs61998192 Chr.7: 105567285-105567285 on Build GRCh38
rs895052984 Chr.7: 105567190-105567190 on Build GRCh38
rs543042477 Chr.7: 105567303-105567303 on Build GRCh38
rs375342336 Chr.7: 105567331-105567331 on Build GRCh38
rs36058979 Chr.7: 105567302-105567302 on Build GRCh38
rs764177063 Chr.7: 105567193-105567193 on Build GRCh38
rs775706636 Chr.7: 105567299-105567299 on Build GRCh38
rs779364605 Chr.7: 105567287-105567287 on Build GRCh38
rs772323757 Chr.7: 105567160-105567160 on Build GRCh38
rs776832049 Chr.7: 105567312-105567312 on Build GRCh38
rs754448407 Chr.7: 105567250-105567250 on Build GRCh38
rs557533198 Chr.7: 105567213-105567213 on Build GRCh38
rs751025208 Chr.7: 105567319-105567320 on Build GRCh38
rs758224586 Chr.7: 105567228-105567228 on Build GRCh38
rs938481898 Chr.7: 105567194-105567194 on Build GRCh38
rs750282703 Chr.7: 105567221-105567221 on Build GRCh38
rs779874101 Chr.7: 105567245-105567245 on Build GRCh38
rs919181672 Chr.7: 105567187-105567187 on Build GRCh38
rs761778810 Chr.7: 105567328-105567328 on Build GRCh38
rs751165359 Chr.7: 105567246-105567246 on Build GRCh38
rs771473733 Chr.7: 105567280-105567280 on Build GRCh38
rs763845178 Chr.7: 105567290-105567290 on Build GRCh38
rs184378923 Chr.7: 105567266-105567266 on Build GRCh38
rs777795343 Chr.7: 105567248-105567248 on Build GRCh38
rs1056894357 Chr.7: 105567200-105567200 on Build GRCh38
rs199604799 Chr.7: 105567295-105567295 on Build GRCh38
rs758122010 Chr.7: 105567180-105567180 on Build GRCh38
rs146607996 Chr.7: 105567171-105567171 on Build GRCh38
rs761878602 Chr.7: 105567212-105567212 on Build GRCh38
rs780817707 Chr.7: 105567262-105567262 on Build GRCh38
rs747595808 Chr.7: 105567293-105567293 on Build GRCh38
rs34310648 Chr.7: 105567208-105567208 on Build GRCh38
rs13533 Chr.7: 105567332-105567332 on Build GRCh38
rs540318781 Chr.7: 105567270-105567270 on Build GRCh38
rs775653724 Chr.7: 105567182-105567182 on Build GRCh38
rs773233825 Chr.7: 105567334-105567334 on Build GRCh38

View Full Product Details